Mettler Toledo S8 Handheld pH/Ion Meter Fluoride Kit

Representative image only.

Mettler Toledo S8 Handheld pH/Ion Meter Fluoride Kit

Mettler Toledo | Mfr: 30207879| Item: 3590365

No more juggling while multitasking

$2,740.51CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$2,740.51CAD / Each

Buy It Together

Current Item

Mettler Toledo S8 Handheld pH/Ion Meter Fluoride Kit
Mettler Toledo S8 Handheld pH/Ion Meter Fluoride Kit
3590365

$2,740.51CAD / Each
+ + =
Total Price 4316.52 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

perfectION Fluoride electrode, LabX direct pH software, USB cable, electrode holder, stabilizing/mounting base, wrist strap, test report, and 4 AA batteries.

Key Features

  • Single-handed operation frees a hand for other tasks
  • Robust IP67-rated housing protects meter from harsh conditions
  • Enhanced contrast screen is readable in any light condition—even bright sunlight
  • Compatible with InLab® ISM sensors for quick electrode changes without recalibration
  • 1 year warranty

More About this Item

This handheld meter withstands the rigors of busy analytical laboratories, demanding process environments, and unpredictable outdoor applications. The directional keypad with central enter key and intuitive simplified interface allow one hand to set up, calibrate, and measure while the other hand remains free. Waterproof housing resists washdown exposure and inclement weather.

Choose the kit with an electrode and accessories that best fit your application.

For fluoride measurements, the perfectION™ combination ion-selective electrode features the built-in Click & Clear™ reference junction to prevent clogging and provide faster, more stable readings.

1 Year Warranty
Specifications
  • Meter TypePortable
  • Display typeGraphic LCD
  • ConnectionsMicro-USB
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001, 0.01, 0.1
  • pH Accuracy±0.002
  • Buffer RecognitionNIST, DIN, JIS, MERCK, MT USA, MT Europe, JJG 119, Technical, Custom
  • CalibrationUp to 5 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1, 1
  • mV Accuracy±0.1
  • Min ISEDepends on the electrode used: perfectION Fluoride ISE lower limit of detection is 0.02 mg/L
  • Max ISEDepends on the electrode used
  • ISE Resolution3 or 4 digits (auto range)
  • ISE Accuracy±0.5%
  • Min Temperature (° C)-5
  • Max Temperature (° C)130
  • Min Temperature (° F)23
  • Max Temperature (° F)266
  • Temperature Resolution0.1°C
  • Temperature Accuracy±0.2° up to 105°C, ±0.5° above 105°C
  • Temperature compensationAutomatic or manual
  • Data Logging (points)2000 with time and date
  • WaterproofYes
  • Battery4 AA (included)
  • Battery life (hr)250
  • Length (cm)22.2
  • Width (cm)7
  • Height (cm)3.5
  • Width (in)2 3/4
  • Length (in)8 3/4
  • Height (in)1 3/8
  • DescriptionHandheld pH/Ion Meter Fluoride Kit
  • Warranty1 year
Reviews
Q&A