Mettler Toledo MA Series Standard Precision Toploading Balances
Simple and efficient balances for routine everyday weighing
  • Large, easy-to-read LCD hybrid touchscreen simplify operation
  • Electromagnetic Force Compensation (EMFC) weighing cell provides accurate and reliable results
  • Internal calibration ensure accuracy
  • Flexible integration options—connect using USB-A and RS-232
  • Automatically recognizes plugged in peripheral devices
  • Easily identify samples using the sample ID function
  • Overload protection guards the weighing cell against excess weight overload
  • High-quality materials and solid construction provide long service life
The MA series precision toploading balances offer excellent repeatability, readability, and durability, making them perfect for routine tasks. The large, easy-to-read LCD hybrid touchscreen provides on-screen instructions for quick and efficient operation even when wearing gloves. An electromagnetic force compensation (EMFC) weighing cell and internal calibration provide consistent reliable results.
The USB-A and RS-232 interfaces allow easy transfer of data to a printer, computer, or USB storage device for complete and error-free documentation. Ten built-in applications include essential weighing functions, statistics, and formulation. Perform weighing operations below the balance with the included weighing hook. Password protection feature restricts modification of the balance settings to authorized personnel only.
Rugged housing, made of high-quality and durable materials, protects the sensitive sensor from environmental influences and impacts. Smooth surfaces and rounded edges provide quick and easy cleaning. Built-in overload protection prolongs the life of your balance. Convenient power-saving mode minimizes energy usage and saves money.
$3,064.65 - $13,275.24CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Mettler Toledo MA103 Precision Toploading Balance, MA Series, 120g x 1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262917 Mfr # 30697420
COMPARE
120 1 Precision Toploading Balance, 120 g x 1 mg with Draft Shield
$3,270.93CAD / Each
Mettler Toledo MA203 Precision Toploading Balance, MA Series, 220g x 1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262918 Mfr # 30697423
COMPARE
220 1 Precision Toploading Balance, 220 g x 1 mg with Draft Shield
$3,536.14CAD / Each
Mettler Toledo MA303 Precision Toploading Balance, MA Series, 320g x 1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262919 Mfr # 30697426
COMPARE
320 1 Precision Toploading Balance, 320 g x 1 mg with Draft Shield
$3,786.61CAD / Each
Mettler Toledo MA503 Precision Toploading Balance, MA Series, 520g x 1mg with LCD Hybrid Touchscreen, Draft Shield, and Internal Calibration 0262920 Mfr # 30697429
COMPARE
520 1 Precision Toploading Balance, 520 g x 1 mg with Draft Shield
$4,353.87CAD / Each
Mettler Toledo MA602 Precision Toploading Balance, MA Series, 620g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262921 Mfr # 30697440
COMPARE
620 10 Precision Toploading Balance, 620 g x 10 mg
$3,064.65CAD / Each
Mettler Toledo MA1002 Precision Toploading Balance, MA Series, 1200g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262922 Mfr # 30697443
COMPARE
1200 10 Precision Toploading Balance, 1200 g x 10 mg
$3,550.87CAD / Each
Mettler Toledo MA2002 Precision Toploading Balance, MA Series, 2200g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262923 Mfr # 30697446
COMPARE
2200 10 Precision Toploading Balance, 2200 g x 10 mg
$3,963.42CAD / Each
Mettler Toledo MA3002 Precision Toploading Balance, MA Series, 3200g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262924 Mfr # 30697449
COMPARE
3200 10 Precision Toploading Balance, 3200 g x 10 mg
$4,066.56CAD / Each
Mettler Toledo MA4002 Precision Toploading Balance, MA Series, 4200g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262925 Mfr # 30697452
COMPARE
4200 10 Precision Toploading Balance, 4200 g x 10 mg
$4,317.03CAD / Each
Mettler Toledo MA6002 Precision Toploading Balance, MA Series, 6200g x 10mg with LCD Hybrid Touchscreen and Internal Calibration 0262926 Mfr # 30697455
COMPARE
6200 10 Precision Toploading Balance, 6200 g x 10 mg
$4,736.95CAD / Each
Mettler Toledo MA5001 Precision Toploading Balance, MA Series, 5200g x 100mg with LCD Hybrid Touchscreen and Internal Calibration 0262927 Mfr # 30697458
COMPARE
5200 100 Precision Toploading Balance, 5200 g x 100 mg
$3,388.80CAD / Each
Mettler Toledo MA12001L Precision Toploading Balance, MA Series, 12.2kg x 100mg with LCD Hybrid Touchscreen and Internal Calibration 0262928 Mfr # 30697475
COMPARE
12200 100 Precision Toploading Balance, 12.2 kg x 100 mg
$11,197.76CAD / Each
Mettler Toledo MA16001L Precision Toploading Balance, MA Series, 16.2kg x 100mg with LCD Hybrid Touchscreen and Internal Calibration 0262929 Mfr # 30697478
COMPARE
16200 100 Precision Toploading Balance, 16.2 kg x 100 mg
$12,214.40CAD / Each
Mettler Toledo MA32001L Precision Toploading Balance, MA Series, 32.2kg x 100mg with LCD Hybrid Touchscreen and Internal Calibration 0262930 Mfr # 30697481
COMPARE
32200 100 Precision Toploading Balance, 32.2 kg x 100 mg
$13,275.24CAD / Each
Mettler Toledo MA32000L Precision Toploading Balance, MA Series, 32.2kg x 1g with LCD Hybrid Touchscreen and Internal Calibration 0262931 Mfr # 30697484
COMPARE
32200 1000 Precision Toploading Balance, 32.2 kg x 1 g
$10,777.85CAD / Each
Showing 1- 15 of 15