Memmert General-Purpose Gravity Convection Ovens
Precise drying and heating in research, science, industry and quality assurance.
  • Quick and intuitive operation through the unique ControlCOCKPIT
  • AtmoCONTROL software provides simple drag-and-drop programming and documentation
These natural convection ovens are perfect for heating and drying applications in research or industrial environments. Featuring the Memmert ControlCOCKPIT for quick and intuative control with simple touch, turn and go operation. TwinDISPLAY gives the user control of device specific parameters and temperature monitoring and graphical display of program sequence. Versatile menu functions make setting of language, time zone, daylight savings, and choice between Celsius and Fahrenheit simple and easy. Set point "wait "function guarantees process time does not start until set temperature is reached. Included USB port allow you to upload programs, read out protocol logs, and protect oven with individual user ID functions.
A built-in overtemperature protection system automatically switches off oven at approximately 10°C above the set temperature with audible and visual alarms. Individual min/max values can be set for the over/under temperature alarm. Other features include as internal data logger with a storage capacity of 10 years and an Ethernet interface for uploading and implementing programs and for online logging.
Durable construction with a scratch-resistant structured stainless steel exterior and easy-to-clean corrosion-resistant stainless steel interior. Recessed grips make oven easy to carry and handle and stainless steel door can be changed from left to ride side opening.
$6,369.46 - $10,875.09CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Memmert UN 75 PlUS 115 VOLT Universal Gravity Convection Oven, 2.7 Cuft, 115V 5220011 Mfr # UN 75 PlUS 115 VOLT
COMPARE
2.7 115
$6,369.46CAD / Each
Memmert UN 110 PLUS 115 VOLT Universal Gravity Convection Oven, 3.8 Cuft, 115V 5220013 Mfr # UN 110 PLUS 115 VOLT
COMPARE
3.8 115
$7,305.07CAD / Each
Memmert UN 260 Plus 230 Volt Universal Gravity Convection Oven, 9 Cuft, 230V 5220017 Mfr # UN 260 Plus 230 Volt
COMPARE
9 230
$10,875.09CAD / Each