MadgeTech Temperature and Humidity Data Loggers
  • Real-time operation
  • Low cost
  • User-friendly
  • Battery powered - user replaceable
  • Programmable start time
  • Reusable
  • Small size
  • Non-volatile solid state memory
  • Readings selectable 1 every 2 sec to 1 every 12 hours
The MH1000 series data loggers can be started and stopped directly from a personal computer and its small size allows it to fit almost anywhere. Data retrieval is quick and easy. Simply plug a logger into an empty COM port and our user-friendly software allows data to be examined, exported and printed.
$674.08 - $710.91CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
MadgeTech RHTEMP1000 Temp/RH Recorder RK-18003-42 Mfr # RHTEMP1000
COMPARE
176
$674.08CAD / Each
MadgeTech RHTEMP1000IS Intrinsically Safe Temp/RH Recorder RK-18003-43 Mfr # RHTEMP1000IS
COMPARE
176
$710.91CAD / Each