MadgeTech RHTEMP1000IS Intrinsically Safe Temp/RH Recorder
Representative image only.
MadgeTech RHTEMP1000IS Intrinsically Safe Temp/RH Recorder
MadgeTech | Mfr: RHTEMP1000IS| Item: 1800343
$800.05CAD / Each
$800.05CAD / Each
Product Description
Key Features
  • Real-time operation
  • Low cost
  • User-friendly
  • Battery powered - user replaceable
  • Programmable start time
  • Reusable
  • Small size
  • Non-volatile solid state memory
  • Readings selectable 1 every 2 sec to 1 every 12 hours
  • 1 year warranty
More About this Item
The MH1000 series data loggers can be started and stopped directly from a personal computer and its small size allows it to fit almost anywhere. Data retrieval is quick and easy. Simply plug a logger into an empty COM port and our user-friendly software allows data to be examined, exported and printed.
1 Year Warranty
Specifications
  • Number of Channels1
  • Input TypeTemperature and humidity
  • Max Temperature (° F)176
  • Min Temperature (° F)-40
  • Max Temperature (° C)80
  • Min Temperature (° C)-40
  • DescriptionIntrinsically Safe Submersible Temp/RH Recorder