IKA Orbital Shakers
Compact – suitable for small containers and microplates
  • Low-profile design ensures stability at high speeds
  • Various attachments provide excellent versatility
  • Timer with countdown function on digital model
These orbital shakers feature continuous or touch operation, triggered by pressing the attachment. The digital model has a 7-segment LED display and timer. Knobbed feet provide stability on the work surface when operating at high speeds. Sturdy zinc die-cast casing.
Electronic speed control can be adjusted from 0 to 3000 rpm. Operate in either mode A or B. Mode A (''safe'' mode) only allows maximum speed to be reached when using the standard attachment, all other attachments are limited to a speed of 1300 rpm. Mode B allows all attachments to operate at maximum speed.
$1,331.94 - $3,788.09CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Operating Temperature (° F) Max Speed (rpm) Power (VAC)
Availability Pricing
IKA MTS 2/4 Orbital Shaker, digital, 0 to 1100 rpm, microtiter plates 0430332 Mfr # 3208001
COMPARE
41 1100 115
$3,788.09CAD / Each
IKA 3617000 MS 3 Basic Orbital Shaker, 0 to 3000 rpm, 100 to 240 VAC 0430400 Mfr # 0003617000
COMPARE
40 3000 100 to 240
$1,331.94CAD / Each
IKA 3319000 MS 3 Digital Orbital Shaker, 0 to 3000 rpm, 100 to 240 VAC 0430404 Mfr # 3319000
COMPARE
40 3000 100 to 240
$1,504.33CAD / Each