Idex® UNF(M) Thread to Barbed Adapters
Easily connect soft-wall tubing to 10-32 coned, 1/4-28 flat-bottom, or 5/16-24 flat-bottom port
  • Choose from ETFE or PEEK construction
Slip soft-wall tubing over the barbed end then thread the adapter into a receiving port for a reliable low-pressure connection.
$6.78 - $48.92CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Compression Size / Tube Od (Port 1) (in) Thread size UNF(M) (port 1) (InchShort)
Availability Pricing
Idex Adapter Fitting, Natural ETFE, 0.040" Bore, 1/16" Hose Barb x Male 1/4-28 Flat Bottom; 1/EA 0201408 Mfr # P-646
COMPARE
1/16 1/16 1/4-28 Flat Bottom
$7.37CAD / Each
Idex Adapter Fitting, Natural ETFE, 0.080" Bore, 1/8" Hose Barb x Male 1/4-28 Flat Bottom; 1/EA 0201409 Mfr # P-647
COMPARE
1/8 1/8 1/4-28 Flat Bottom
$7.37CAD / Each
Idex Adapter Fitting, Natural ETFE, 0.040" Bore, 1/16" Hose Barb x Male 10-32 Coned; 1/EA 0201417 Mfr # P-663
COMPARE
1/16 1/16 10-32 Coned
$7.22CAD / Each
Idex Adapter Fitting, Natural ETFE, 0.040" Bore, 1/8" Hose Barb x Male 10-32 Coned; 1/EA 0201418 Mfr # P-664
COMPARE
1/8 1/8 10-32 Coned
$7.22CAD / Each
Idex Adapter Fitting, Natural PEEK, 0.040" Bore, 1/16" Hose Barb x Male 1/4-28 Flat Bottom; 1/EA 0201419 Mfr # P-668
COMPARE
1/16 1/16 1/4-28 Flat Bottom
$16.35CAD / Each
Idex Adapter Fitting, Natural ETFE, 0.10" Bore, 3/16" Hose Barb x 1/4-28 Flat Bottom; 1/EA 0202355 Mfr # P-648
COMPARE
3/16 3/16 1/4-28 Flat Bottom
$6.78CAD / Each
Idex Adapter Fitting, Natural ETFE, 0.10" Bore, 3/16" Hose Barb x 5/16-24 Flat Bottom; 1/EA 0202367 Mfr # P-689
COMPARE
3/16 3/16 5/16-24 Flat Bottom
$48.92CAD / Each
Idex Adapter Fitting, for P-799, Natural ETFE, 0.060" Bore, 3/32 to 1/8" Hose Barb x Male 1/4-28 Flat Bottom; 1/EA 0202368 Mfr # P-691
COMPARE
1/8 3/32 1/4-28 Flat Bottom
$12.82CAD / Each
Idex Adapter Fitting, for P-794, Natural PEEK, 0.020" Bore, 0.020" to 0.030" Hose Barb x Male 1/4-28 Flat Bottom; 1/EA 0202369 Mfr # P-692
COMPARE
1/32 3/32 1/4-28 Flat Bottom
$24.90CAD / Each
Showing 1- 9 of 9