Idex® Low-Pressure Bulkhead Unions
Thread through the equipment housingto connect the external system to tubing
  • Manufactured from inert polymers
Use bulkhead unions in a variety of low-pressure situations throughout your fluidic system. Each PEEK union has unique 3/8-24 external threads and can be held in place with included stainless steel nuts and lock washers. Requires 3/8" hole to mount. Assemblies include nut and washer.
$25.05 - $36.98CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Thread size UNF(F) (port 1) (InchShort) Description
Availability Pricing
Idex Low-Pressure Union Assembly, Bulkhead, Natural PEEK, 0.020" Bore, 10-32 Coned; 1/EA 0201387 Mfr # P-440
COMPARE
PEEK 10-32 Coned Low-Pressure Union Assembly, Bulkhead, Natural PEEK, 0.020" Bore, 10-32 Coned; 1/EA
$29.62CAD / Each
Idex Low-Pressure Union Assembly, Bulkhead, Red PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA 0201388 Mfr # P-441
COMPARE
PEEK 1/4-28 Flat Bottom Low-Pressure Union Assembly, Bulkhead, Red PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA
$28.58CAD / Each
Idex Low-Pressure Union Assembly, Bulkhead, Natural PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA 0201389 Mfr # P-441N
COMPARE
PEEK 1/4-28 Flat Bottom Low-Pressure Union Assembly, Bulkhead, Natural PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA
$28.58CAD / Each
Idex Low-Pressure Union Body, Bulkhead, Natural PEEK, 0.020" Bore, 10-32 Coned; 1/EA 0202322 Mfr # P-440-01
COMPARE
PEEK 10-32 Coned Low-Pressure Union Body, Bulkhead, Natural PEEK, 0.020" Bore, 10-32 Coned; 1/EA
$25.05CAD / Each
Idex Low-Pressure Union Body, Bulkhead, Red PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA 0202323 Mfr # P-441-01
COMPARE
PEEK 1/4-28 Flat Bottom Low-Pressure Union Body, Bulkhead, Red PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA
$36.98CAD / Each
Idex Low-Pressure Union Body, Bulkhead, Black PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA 0202324 Mfr # P-441-01BLK
COMPARE
PEEK 1/4-28 Flat Bottom Low-Pressure Union Body, Bulkhead, Black PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA
$36.98CAD / Each
Idex Low-Pressure Union Body, Bulkhead, Natural PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA 0202325 Mfr # P-441-01N
COMPARE
PEEK 1/4-28 Flat Bottom Low-Pressure Union Body, Bulkhead, Natural PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA
$25.05CAD / Each
Idex Low-Pressure Union Assembly, Bulkhead, Blue PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA 0202326 Mfr # P-441B
COMPARE
PEEK 1/4-28 Flat Bottom Low-Pressure Union Assembly, Bulkhead, Blue PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA
$28.58CAD / Each
Idex Low-Pressure Union Assembly, Bulkhead, Black PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA 0202327 Mfr # P-441BLK
COMPARE
PEEK 1/4-28 Flat Bottom Low-Pressure Union Assembly, Bulkhead, Black PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA
$28.58CAD / Each
Idex Low-Pressure Union Assembly, Bulkhead, Green PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA 0202328 Mfr # P-441G
COMPARE
PEEK 1/4-28 Flat Bottom Low-Pressure Union Assembly, Bulkhead, Green PEEK, 0.040" Bore, 1/4-28 Flat Bottom; 1/EA
$28.58CAD / Each
Showing 1- 10 of 10