Idex® Inlet Solvent Filters
Keep damaging particles out of your system with inlet solvent filters
  • Protect your system from particulate matter that can cause damage
  • Filters available to fit specific system specifications
  • Easy to replace without tools
For good preventive maintenance, inlet solvent filters are an optimal way to cleanse solvents, avoiding pump cavitation and system damage. Different style stainless steel filters are available for specific system specifications and can be easily replaced regularly without tools.
$35.36 - $432.44CAD / Each
23 variations of this product are available.
See All Product Options Below
23 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Material Pore Size (µm) Description
Availability Pricing
Idex Inlet Solvent Filter, Stem, SS, 10 µm, 1/16" ID Tubing; 1/EA 4270100 Mfr # A-302
COMPARE
Stainless Steel 10 Inlet Solvent Filter, Stem, SS, 10 µm, 1/16" ID Tubing; 1/EA
$50.54CAD / Each
Idex Inlet Solvent Filter, Stem, SS, PCTFE Flangeless Fitting, 10 µm, 1/8" OD Tubing; 1/EA 4270102 Mfr # A-302A
COMPARE
Stainless Steel 10 Inlet Solvent Filter, Stem, SS, PCTFE Flangeless Fitting, 10 µm, 1/8" OD Tubing; 1/EA
$87.81CAD / Each
Idex Inlet Solvent Filter Kit, SS/PCTFE, 10 µm, 1/8" OD Tubing; 1/EA 4271100 Mfr # A-200
COMPARE
Stainless Steel 10 Inlet Solvent Filter Kit, SS/PCTFE, 10 µm, 1/8" OD Tubing; 1/EA
$331.51CAD / Each
Idex Inlet Solvent Filter Kit, SS/PCTFE, 2 µm, 1/4" OD Tubing; 1/EA 4271101 Mfr # A-201
COMPARE
Stainless Steel 2 Inlet Solvent Filter Kit, SS/PCTFE, 2 µm, 1/4" OD Tubing; 1/EA
$201.12CAD / Each
Idex Inlet Solvent Filter, SS, 10 µm, 1/8" OD Tubing; 1/EA 4271102 Mfr # A-220
COMPARE
Stainless Steel 10 Inlet Solvent Filter, SS, 10 µm, 1/8" OD Tubing; 1/EA
$37.28CAD / Each
Idex Inlet Solvent Filter, SS, 10 µm, 1/8" OD Tubing; 5/PK 4271103 Mfr # A-221
COMPARE
Stainless Steel 10 Inlet Solvent Filter, SS, 10 µm, 1/8" OD Tubing; 5/PK
$182.55CAD / Pkg of 5
Idex Inlet Solvent Filter, SS, 2 µm, 1/8" OD Tubing; 1/EA 4271104 Mfr # A-222
COMPARE
Stainless Steel 2 Inlet Solvent Filter, SS, 2 µm, 1/8" OD Tubing; 1/EA
$35.36CAD / Each
Idex Inlet Solvent Filter, SS, 2 µm, 1/8" OD Tubing; 5/PK 4271105 Mfr # A-223
COMPARE
Stainless Steel 2 Inlet Solvent Filter, SS, 2 µm, 1/8" OD Tubing; 5/PK
$160.89CAD / Pkg of 5
Idex Inlet Solvent Filter, Stem, SS, 20 µm, 1/16" ID Tubing; 1/EA 4271106 Mfr # A-225
COMPARE
Stainless Steel 20 Inlet Solvent Filter, Stem, SS, 20 µm, 1/16" ID Tubing; 1/EA
$49.36CAD / Each
Idex lnlet Solvent Filter, SS/PCTFE, 20 µm, 1/8" OD Tubing; 1/EA 4271107 Mfr # A-225A
COMPARE
Stainless Steel 20 lnlet Solvent Filter, SS/PCTFE, 20 µm, 1/8" OD Tubing; 1/EA
$71.46CAD / Each
Idex lnlet Solvent Filter, SS, 20 µm, 1/8" OD Tubing; 1/EA 4271108 Mfr # A-225A-1
COMPARE
Stainless Steel 20 lnlet Solvent Filter, SS, 20 µm, 1/8" OD Tubing; 1/EA
$106.82CAD / Each
Idex lnlet Solvent Filter, SS/PCTFE, 10 µm, 5/16" OD Tubing; 1/EA 4271109 Mfr # A-226A
COMPARE
Stainless Steel 10 lnlet Solvent Filter, SS/PCTFE, 10 µm, 5/16" OD Tubing; 1/EA
$432.44CAD / Each
Idex lnlet Solvent Filter, SS/PCTFE, 10 µm, 1/4" OD Tubing; 1/EA 4271110 Mfr # A-227A
COMPARE
Stainless Steel 10 lnlet Solvent Filter, SS/PCTFE, 10 µm, 1/4" OD Tubing; 1/EA
$149.25CAD / Each
Idex lnlet Solvent Filter, SS, 20 µm, 1/4" OD Tubing; 1/EA 4271112 Mfr # A-230A
COMPARE
Stainless Steel 20 lnlet Solvent Filter, SS, 20 µm, 1/4" OD Tubing; 1/EA
$156.33CAD / Each
Idex lnlet Solvent Filter, SS/PCTFE, 20 µm, 3/16" OD Tubing; 1/EA 4271113 Mfr # A-231A
COMPARE
Stainless Steel 20 lnlet Solvent Filter, SS/PCTFE, 20 µm, 3/16" OD Tubing; 1/EA
$72.93CAD / Each
Idex Inlet Solvent Filter, SS/PCTFE, 10 µm, 1/8" OD Tubing; 5/PK 4271115 Mfr # A-241
COMPARE
Stainless Steel 10 Inlet Solvent Filter, SS/PCTFE, 10 µm, 1/8" OD Tubing; 5/PK
$318.25CAD / Pkg of 5
Idex Inlet Solvent Filter, SS/PCTFE, 2 µm, 1/8" OD Tubing; 1/EA 4271116 Mfr # A-242
COMPARE
Stainless Steel 2 Inlet Solvent Filter, SS/PCTFE, 2 µm, 1/8" OD Tubing; 1/EA
$53.63CAD / Each
Idex Inlet Solvent Filter, SS/PCTFE, 2 µm, 1/8" OD Tubing; 5/PK 4271117 Mfr # A-243
COMPARE
Stainless Steel 2 Inlet Solvent Filter, SS/PCTFE, 2 µm, 1/8" OD Tubing; 5/PK
$351.40CAD / Pkg of 5
Idex Inlet Solvent Filter, Stem, Precleaned, SS, 10 µm, 1/16" ID Tubing; 1/EA 4271118 Mfr # A-302S
COMPARE
Stainless Steel 10 Inlet Solvent Filter, Stem, SS, 10 µm, 1/16" ID Tubing; 1/EA
$95.33CAD / Each
Idex Inlet Solvent Filter Assembly, Stem, Bottom-of-the-Bottle, SS, 10 µm, 1/16" ID Tubing; 1/EA 4271119 Mfr # A-309
COMPARE
Stainless Steel 10 Inlet Solvent Filter Assembly, Stem, Bottom-of-the-Bottle, SS, 10 µm, 1/16" ID Tubing; 1/EA
$54.66CAD / Each
Showing 1 - 20 of 23