Idex® High-Pressure PEEK ZDV Unions
PEEK ZDV unions provide accurate results
  • Inert PEEK fluid path
  • 10-32 coned configuration
These Idex® high-pressure PEEK ZDV unions provide a tight, secure connection in your chromatography fluid path.
$37.57 - $57.76CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Idex High-Pressure Union Assembly, ZDV, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0201429 Mfr # P-704
COMPARE
1/16 PEEK High-Pressure Union Assembly, ZDV, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA
$57.46CAD / Each
Idex High-Pressure Union Assembly, ZDV, PEEK, 0.010" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0201437 Mfr # P-742
COMPARE
1/16 PEEK High-Pressure Union Assembly, ZDV, PEEK, 0.010" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA
$57.46CAD / Each
Idex High-Pressure Union Assembly, ZDV, PEEK, 0.050" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0201440 Mfr # P-760
COMPARE
1/16 PEEK High-Pressure Union Assembly, ZDV, PEEK, 0.050" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA
$57.76CAD / Each
Idex High-Pressure Union Body, True ZDV, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202449 Mfr # P-704-01
COMPARE
1/16 PEEK High-Pressure Union Body, True ZDV, PEEK, 0.020" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA
$37.57CAD / Each
Idex High-Pressure Union Body, True ZDV, PEEK, 0.010" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202459 Mfr # P-742-01
COMPARE
1/16 PEEK High-Pressure Union Body, True ZDV, PEEK, 0.010" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA
$38.75CAD / Each
Idex High-Pressure Union Body, True ZDV, PEEK, 0.050" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA 0202460 Mfr # P-760-01
COMPARE
1/16 PEEK High-Pressure Union Body, True ZDV, PEEK, 0.050" ID, 1/16" OD Tubing, 10-32 Coned; 1/EA
$38.90CAD / Each