Idex® High-Pressure MicroTight® Adapters for 1/16" OD to 360 µm OD Tubing

Create a high-pressure connection between 1/16" OD tubing and capillary tubing
  • Securely join together fluid pathway components
  • Manufactured from 316 stainless steel or inert polymers
  • Holds tight connection even through vibration
Port 1 is 10-32 coned for 1/16" OD tubing.
$66.89 - $407.39CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Thread size UNF(F) (port 2) (InchShort) Description
Availability Pricing
Idex MicroTight Adapter Assy, True ZDV, Blue PEEK, 1/16" to 360 µm OD, 10-32 to 6-32 0201460 Mfr # P-882
COMPARE
PEEK 6-32 Coned MicroTight Adapter Assembly, True ZDV, Blue PEEK, 1/16" to 360 µm OD Tubing, 10-32 Coned to 6-32 Coned; 1/EA
$66.89CAD / Each
Idex Ultra High-Pressure MicroAdapter Assy, SS, 1/16" to 360 µm OD, 10-32 to 6-32 0202512 Mfr # UH-634
COMPARE
Stainless Steel 6-32 Coned Ultra High-Pressure MicroTight® Adapter Assembly, SS, 1/16" to 360 µm OD Tubing, 10-32 to 6-32 Coned; 1/EA
$110.95CAD / Each
Idex Ultra High-Pressure MicroTee Assy, SS, Blk, 1/16" to 360 µm OD, 10-32 to 5/16-24 0202515 Mfr # UH-753
COMPARE
SS/PEEK 5/16-24 Coned Ultra High-Pressure MicroTee Assembly, SS, Black, 1/16" to 360 µm OD Tubing, 10-32 to 5/16-24 Coned; 1/EA
$407.39CAD / Each
Idex Ultra High-Pressure MicroCross Assy, SS, 1/16" to 360 µm OD, 10-32 to 5/16-24 0202516 Mfr # UH-906
COMPARE
Stainless Steel 5/16-24 Coned Ultra High-Pressure MicroCross Assembly, SS, 1/16" to 360 µm OD Tubing, 10-32 to 5/16-24 Coned; 1/EA
$341.83CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat