Idex® Flangeless FlushNut™ Fitting Assemblies, 1/4-28 Flat-Bottom Connection
FlushNut nuts are perfect for tight-space applications
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
$21.81 - $213.79CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Thread size UNF(M) (port 2) (InchShort)
Availability Pricing
Idex Flangeless Fitting, FlushNut™, SS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202052 Mfr # XF-358
COMPARE
1/16 1/4-28 flat bottom
$23.43CAD / Each
Idex Flangeless Fitting, FlushNut™, SS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0202053 Mfr # XF-358X
COMPARE
1/16 1/4-28 flat bottom
$213.79CAD / Pkg of 10
Idex Flangeless Fitting, FlushNut™, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom, 1/EA 0202054 Mfr # XF-368
COMPARE
1/8 1/4-28 flat bottom
$21.81CAD / Each