Grinding Balls for Retsch Mills
Zirconium grinding balls offer excellent durability
In addition to the instrument settings, the filling level of the jar is crucial for the success of the grinding process in mixer jar mills. A jar filling should consist of about 1⁄3 sample and 1⁄3 ball charge (pieces). The remaining third is the free jar volume that is necessary for the free movement of the balls. Exceptions to this are voluminous materials, such as wool, leaves, and grasses. In these cases a material filling level of 70 to 80% is necessary.
$5.45 - $277.00CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Retsch 05.368.0034 Stainless Steel Grinding Ball for Jar Mills, 5 mm; 1/Pk 0418259 Mfr # 05.368.0034
COMPARE
Stainless Steel Grinding Ball for Jar Mills, 5 mm; 1/Pk
$5.45CAD / Each
Retsch 05.368.0035 Stainless Steel Grinding Ball for Jar Mills, 7 mm; 1/Pk 0418260 Mfr # 05.368.0035
COMPARE
Stainless Steel Grinding Ball for Jar Mills, 7 mm; 1/Pk
$5.89CAD / Each
Retsch 05.368.0036 Stainless Steel Grinding Ball for Jar Mills, 9 mm; 1/Pk 0418261 Mfr # 05.368.0036
COMPARE
Stainless Steel Grinding Ball for Jar Mills, 9 mm; 1/Pk
$7.51CAD / Each
Retsch 05.368.0037 Stainless Steel Grinding Ball for Jar Mills, 12 mm; 1/Pk 0418262 Mfr # 05.368.0037
COMPARE
Stainless Steel Grinding Ball for Jar Mills, 12 mm; 1/Pk
$12.38CAD / Each
Retsch 05.368.0062 Stainless Steel Grinding Ball for Jar Mills, 20 mm; 1/Pk 0418263 Mfr # 05.368.0062
COMPARE
Stainless Steel Grinding Ball for Jar Mills, 20 mm; 1/Pk
$39.93CAD / Each
Retsch 05.368.0024 Agate Grinding Ball for Jar Mills, 5 mm; 1/Pk 0418264 Mfr # 05.368.0024
COMPARE
Agate Grinding Ball for Jar Mills, 5 mm; 1/Pk
$20.19CAD / Each
Retsch 05.368.0025 Agate Grinding Ball for Jar Mills, 7 mm; 1/Pk 0418265 Mfr # 05.368.0025
COMPARE
Agate Grinding Ball for Jar Mills, 7 mm; 1/Pk
$23.72CAD / Each
Retsch 05.368.0026 Agate Grinding Ball for Jar Mills, 9 mm; 1/Pk 0418266 Mfr # 05.368.0026
COMPARE
Agate Grinding Ball for Jar Mills, 9 mm; 1/Pk
$28.73CAD / Each
Retsch 05.368.0096 Grinding Ball, Zirconium, 12 mm Dia 0418272 Mfr # 05.368.0096
COMPARE
Grinding Ball, Zirconium, 12-mm
$152.86CAD / Each
Retsch 05.368.0113 Grinding Ball, Zirconium, 15 mm Dia 0418273 Mfr # 05.368.0113
COMPARE
Grinding Ball, Zirconium, 15-mm
$277.00CAD / Each
Retsch 05.368.0105 Stainless steel grinding balls, 25 mm 25 mm 0418565 Mfr # 05.368.0105
COMPARE
Stainless steel grinding balls, 25 mm 25 mm
$75.14CAD / Each
Showing 1- 11 of 11