GHS Environment Pictogram Labels

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Environment pictogram indicates the following potential hazard: aquatic toxicity.
$28.44 - $87.22CAD / Roll of 250

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Quantity
Availability Pricing
Accuform LZH608PS2 GHS Label, Environment Pictogram, 1" x 1", adhesive paper, 250/roll 5071308 Mfr # LZH608PS2
COMPARE
1 Adhesive Paper 250/roll
$28.44CAD / Roll of 250
Accuform LZH608PS5 GHS Label, Environment Pictogram, 1" x 1", adhesive paper, 500/roll 5071309 Mfr # LZH608PS5
COMPARE
1 Adhesive Paper 500/roll
$46.26CAD / Roll of 500
Accuform LZH608EV2 GHS Label, Environment Pictogram, 1" x 1", adhesive poly, 250/roll 5071310 Mfr # LZH608EV2
COMPARE
1 Adhesive Poly 250/roll
$43.91CAD / Roll of 250
Accuform LZH618EV5 GHS Label, Environment Pictogram, 2" x 2", adhesive poly, 500/roll 5071315 Mfr # LZH618EV5
COMPARE
2 Adhesive Poly 500/roll
$87.22CAD / Roll of 500
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat