Accuform LZH608PS5 GHS Label, Environment Pictogram, 1" x 1", adhesive paper, 500/roll

Representative image only.

Accuform LZH608PS5 GHS Label, Environment Pictogram, 1" x 1", adhesive paper, 500/roll

Accuform | Mfr: LZH608PS5| Item: 5071309

$46.26CAD / Roll of 500

$46.26CAD / Roll of 500
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Environment pictogram indicates the following potential hazard: aquatic toxicity.

Specifications
  • HeaderENVIRONMENT
  • LanguagesEnglish
  • MaterialAdhesive Paper
  • Width (in)1
  • Height (in)1
  • Quantity500/roll
  • DescriptionGHS Label, Environment Pictogram, 1" x 1", adhesive paper, 500/roll
Reviews
Q&A