Accuform LZH608EV2 GHS Label, Environment Pictogram, 1" x 1", adhesive poly, 250/roll

Representative image only.

Accuform LZH608EV2 GHS Label, Environment Pictogram, 1" x 1", adhesive poly, 250/roll

Accuform | Mfr: LZH608EV2| Item: 5071310

$43.91CAD / Roll of 250

$43.91CAD / Roll of 250
Product Description

More About this Item

GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Environment pictogram indicates the following potential hazard: aquatic toxicity.

Specifications
  • HeaderENVIRONMENT
  • LanguagesEnglish
  • MaterialAdhesive Poly
  • Width (in)1
  • Height (in)1
  • Quantity250/roll
  • DescriptionGHS Label, Environment Pictogram, 1" x 1", adhesive poly, 250/roll
Reviews
Q&A