Fluke® Solmetric High-Efficiency PV Analyzer I-V Curve Tracer Kits
Optimize solar panel performance with high-efficiency PV analyzer I-V curve tracer kits
  • Accurately assess the health and performance of solar photovoltaic (PV) modules and arrays
  • Uses I-V (current versus voltage) curve tracing for quick and detailed performance data
  • Pinpoint problems early to maximize solar energy production while minimizing downtime
  • Test up to 1500 V and up to 30 A
  • Sweep-to-sweep delay of 9 seconds to measure 3.5 MW within an hour
  • Intuitive user interface for easy navigation and real-time analysis
  • SolSensor™ PV reference sensor measures irradiance and temperature
  • Take measurements even in hot environments
  • Wireless connection helps with quick setup and mobility
Enhance the output and prolong the service life of solar modules and arrays by accurately assessing the health and performance with the PV analyzer kits. Select from four kits that meet your application needs. Each kit includes a PVA-1500HE2 analyzer I-V curve tracers and additional Fluke meter(s) to help maximize efficiency and optimize PV performance. Download the data sheet for each instrument for complete specifications.
The high-efficiency PV analyzer is ideal for testing high-efficiency modules (>19% module efficiency). The high-efficiency modules have a large in-rush current when you start the test that can overheat the test instrument and slow down testing. Uniquely designed high-efficiency PV analyzer measures all string types, including high-efficiency modules, up to 30 A. These high-precision analyzers use I-V curve tracing, the most complete solar module performance measurement, to quickly obtain detailed performance data of PV modules.
Measure irradiance and module temperature by clamping the wireless SolSensor™ PV reference sensor (included with analyzer) to the module frame. With the angular response of each unit calibrated for rotation and elevation, the temperature-compensated sensor offers excellent accuracy over a broad range of technologies, sky conditions, and sun angles.
A portable design and an intuitive interface make it easy to use the analyzer in the field. The PV analyzer and the SolSensor sensor communicate wirelessly with a tablet or a laptop (Windows® only) via Wi-Fi. Download the Data Analysis Tool software to display the data in multiple, easy-to-read formats and a statistical overview of the entire array. The software looks at all the collected data for comparison to see where there are differences, identifying the issue for further troubleshooting. Easily create and send customized reports.
$27,369.68 - $35,418.81CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
DC Max (amps) DC Max (Volts) Description
Availability Pricing
Fluke High-Efficiency PV Analyzer I-V Curve Tracer and Thermal Imager Kit 1778507 Mfr # FLUKE-SOL-TI-27HZKIT
COMPARE
30 1500 High-Efficiency PV Analyzer I-V Curve Tracer and Thermal Imager Kit
$35,418.81CAD / Each
Fluke High-Efficiency PV Analyzer I-V Curve Tracer, Clamp, and Multimeter Kit 1778508 Mfr # FLUKE-SOL-DMM87V-KIT
COMPARE
30 1500 High-Efficiency PV Analyzer I-V Curve Tracer, Clamp, and Multimeter Kit
$27,369.68CAD / Each
Fluke High-Efficiency PV Analyzer I-V Curve Tracer, Insulation, Clamp, and Multimeter Kit 1778509 Mfr # FLUKE-SOL-INS87-KIT
COMPARE
30 1500 High-Efficiency PV Analyzer I-V Curve Tracer, Insulation, Clamp, and Multimeter Kit
$29,140.69CAD / Each
Fluke High-Efficiency PV Analyzer I-V Curve Tracer and Insulation Multimeter Kit 1778510 Mfr # FLUKE-SOL-INS37-KIT
COMPARE
30 1500 High-Efficiency PV Analyzer I-V Curve Tracer and Insulation Tester Kit
$28,978.62CAD / Each