Fluke High-Efficiency PV Analyzer I-V Curve Tracer and Thermal Imager Kit

Representative image only.

Fluke High-Efficiency PV Analyzer I-V Curve Tracer and Thermal Imager Kit

Fluke | Mfr: FLUKE-SOL-TI-27HZKIT| Item: 1778507

Optimize solar panel performance with high-efficiency PV analyzer I-V curve tracer kits

$35,418.81CAD / Each

$35,418.81CAD / Each
Product Description

Includes

PVA-1500HE2 analyzer: SolSensor sensor, clamp, MC4 wrenches, test lead sets (banana to alligator clip and banana to MC4), two type K thermocouple sensors, 50 adhesive discs for thermocouple attachment, sensor cleaning supplies, charging supplies, soft case, and hard carrying case.
TiS75+ thermal imaging camera: AC power supply (including universal AC adapters, two-bay smart battery charger, two rugged Li-ion smart batteries, USB cable, 4 GB micro SD card, hard carrying case, soft transport bag, and adjustable hand strap.

Key Features

  • Accurately assess the health and performance of solar photovoltaic (PV) modules and arrays
  • Uses I-V (current versus voltage) curve tracing for quick and detailed performance data
  • Pinpoint problems early to maximize solar energy production while minimizing downtime
  • Test up to 1500 V and up to 30 A
  • Sweep-to-sweep delay of 9 seconds to measure 3.5 MW within an hour
  • Intuitive user interface for easy navigation and real-time analysis
  • SolSensor™ PV reference sensor measures irradiance and temperature
  • Take measurements even in hot environments
  • Wireless connection helps with quick setup and mobility
  • Kit includes a PVA-1500HE2 analyzer and TiS75+ thermal imaging camera
  • View thermal images of hotspots and faulty connections on solar panels
  • High-visibility LCD touch screen
  • Asset tagging allows you to easily compare asset images over time
  • Can store up to 500 images on internal storage
  • 1 year warranty

More About this Item

Enhance the output and prolong the service life of solar modules and arrays by accurately assessing the health and performance with the PV analyzer kits. Select from four kits that meet your application needs. Each kit includes a PVA-1500HE2 analyzer I-V curve tracers and additional Fluke meter(s) to help maximize efficiency and optimize PV performance. Download the data sheet for each instrument for complete specifications.

The high-efficiency PV analyzer is ideal for testing high-efficiency modules (>19% module efficiency). The high-efficiency modules have a large in-rush current when you start the test that can overheat the test instrument and slow down testing. Uniquely designed high-efficiency PV analyzer measures all string types, including high-efficiency modules, up to 30 A. These high-precision analyzers use I-V curve tracing, the most complete solar module performance measurement, to quickly obtain detailed performance data of PV modules.

Measure irradiance and module temperature by clamping the wireless SolSensor™ PV reference sensor (included with analyzer) to the module frame. With the angular response of each unit calibrated for rotation and elevation, the temperature-compensated sensor offers excellent accuracy over a broad range of technologies, sky conditions, and sun angles.

A portable design and an intuitive interface make it easy to use the analyzer in the field. The PV analyzer and the SolSensor sensor communicate wirelessly with a tablet or a laptop (Windows® only) via Wi-Fi. Download the Data Analysis Tool software to display the data in multiple, easy-to-read formats and a statistical overview of the entire array. The software looks at all the collected data for comparison to see where there are differences, identifying the issue for further troubleshooting. Easily create and send customized reports.

Use the includes TiS75+ thermal imaging camera to detect anomolies such as hot spots and faulty electrical connections on solar panels. The handheld device features an industrial-quality LCD touch screen that is easy to use. Save the images to internal memory or to a removable card that can be transferred to a PC via a wireless or a USB connection.

Conformite Europeenne1 Year Warranty
Specifications
  • DC Min (Volts)20
  • DC Max (Volts)1500
  • DC Voltage Accuracy±(0.5% ±0.25V)
  • DC Max (amps)30
  • DC Current Accuracy±(0.5% ±0.04A)
  • DisplayTiS75+: 3.5" (8.9 cm landscape) touchscreen, 640 x 480 LCD
  • MemoryTiS75+: approx. 500 images (internal) and ≥4 GB SD card; >1000 images (external )
  • InterfacePVA-1500HE2: Wi-Fi
    TiS75+: Wi-Fi, LAN (where available), and USB
  • Safety RatingPVA-1500HE2: CAT III 1500 V; IEC 61010-1: Pollution Degree 2
    TiS75+: IEC 61010-1: Pollution Degree 2
  • PowerPVA-1500HE2 and SolSensor: rechargeable batteries
    TiS75+: two rechargeable Li-ion battery (included) or AC operation with included power supply.
  • DescriptionHigh-Efficiency PV Analyzer I-V Curve Tracer and Thermal Imager Kit
  • Warranty1 year
Reviews
Q&A