Fluke High-Efficiency PV Analyzer I-V Curve Tracer, Clamp, and Multimeter Kit

Representative image only.

Fluke High-Efficiency PV Analyzer I-V Curve Tracer, Clamp, and Multimeter Kit

Fluke | Mfr: FLUKE-SOL-DMM87V-KIT| Item: 1778508

Optimize solar panel performance with high-efficiency PV analyzer I-V curve tracer kits

$27,369.68CAD / Each

$27,369.68CAD / Each
Product Description

Includes

PVA-1500HE2 analyzer: SolSensor sensor, clamp, MC4 wrenches, test lead sets (banana to alligator clip and banana to MC4), two type K thermocouple sensors, 50 adhesive discs for thermocouple attachment, sensor cleaning supplies, charging supplies, soft case, and hard carrying case.
393 FC clamp meter: test leads (CAT III 1500 V rated, right angle plugs, with safety caps), iFlex 18" flexible current probe, TPAK magnetic hanging strap, and premium carrying case.
87V multimeter: TL75 test leads, AC175 alligator clips, 80BK temperature probe, holster with tilt-leg, test lead storage, and battery.

Key Features

  • Accurately assess the health and performance of solar photovoltaic (PV) modules and arrays
  • Uses I-V (current versus voltage) curve tracing for quick and detailed performance data
  • Pinpoint problems early to maximize solar energy production while minimizing downtime
  • Test up to 1500 V and up to 30 A
  • Sweep-to-sweep delay of 9 seconds to measure 3.5 MW within an hour
  • Intuitive user interface for easy navigation and real-time analysis
  • SolSensor™ PV reference sensor measures irradiance and temperature
  • Take measurements even in hot environments
  • Wireless connection helps with quick setup and mobility
  • Kit includes a PVA-1500HE2 analyzer, 393 FC solar clamp meter, and 87V digital multimeter
  • Take precise DC current and voltage measurements with clamp meter
  • Multimeter enables AC/DC voltage, current, resistance, and capacitance measurements
  • 1 year warranty

More About this Item

Enhance the output and prolong the service life of solar modules and arrays by accurately assessing the health and performance with the PV analyzer kits. Select from four kits that meet your application needs. Each kit includes a PVA-1500HE2 analyzer I-V curve tracers and additional Fluke meter(s) to help maximize efficiency and optimize PV performance. Download the data sheet for each instrument for complete specifications.

The high-efficiency PV analyzer is ideal for testing high-efficiency modules (>19% module efficiency). The high-efficiency modules have a large in-rush current when you start the test that can overheat the test instrument and slow down testing. Uniquely designed high-efficiency PV analyzer measures all string types, including high-efficiency modules, up to 30 A. These high-precision analyzers use I-V curve tracing, the most complete solar module performance measurement, to quickly obtain detailed performance data of PV modules.

Measure irradiance and module temperature by clamping the wireless SolSensor™ PV reference sensor (included with analyzer) to the module frame. With the angular response of each unit calibrated for rotation and elevation, the temperature-compensated sensor offers excellent accuracy over a broad range of technologies, sky conditions, and sun angles.

A portable design and an intuitive interface make it easy to use the analyzer in the field. The PV analyzer and the SolSensor sensor communicate wirelessly with a tablet or a laptop (Windows® only) via Wi-Fi. Download the Data Analysis Tool software to display the data in multiple, easy-to-read formats and a statistical overview of the entire array. The software looks at all the collected data for comparison to see where there are differences, identifying the issue for further troubleshooting. Easily create and send customized reports.

In addition to the PVA1500HE2 analyzer, this kit contains a 393 FC solar clamp meter, and a 87V digital multimeter, offering a complete solution for solar installation diagnostics and maintenance. The solar clamp meter takes precise DC current and voltage measurements, making it easy to test the performance and troubleshoot solar panels and inverters. Use the multimeter for verifying system integrity and diagnosing electrical faults.

Conformite Europeenne1 Year Warranty
Specifications
  • DC Min (Volts)20
  • DC Max (Volts)1500
  • DC Voltage Accuracy±(0.5% ±0.25V)
  • DC Max (amps)30
  • DC Current Accuracy±(0.5% ±0.04A)
  • Display393 FC: dual display with backlight
    87V: digital-6000 counts, updates 4/sec; 19,999 counts in high-resolution mode; analog-32 segments, updates 40/sec
  • InterfacePVA-1500HE2: Wi-Fi
  • Safety RatingPVA-1500HE2: CAT III 1500 V; IEC 61010-1: Pollution Degree 2
    393 FC: IEC 61010-1, Pollution Degree 2; IEC 61010-2-032: CAT III 1500 V / CAT IV 600 V; IEC 61010-2-033: CAT III 1500 V / CAT IV 600 V
    87V: EN 61010–1 to 1000 V CAT III, 600V CAT IV
  • PowerPVA-1500HE2 and SolSensor: rechargeable batteries
    393 FC: 2 AA batteries
    87V: 9 V battery (included)
  • DescriptionHigh-Efficiency PV Analyzer I-V Curve Tracer, Clamp, and Multimeter Kit
  • Warranty1 year
Reviews
Q&A