Fluke Networks LinkIQ™ Network Testers and Kits
Verify cable performance up to 10 Gbps using frequency-based measurements
  • Provides distance to fault information along with a wire map of the cable under test
  • Performs ping tests to verify connectivity
  • Control and view information on the large touchscreen color display
  • Measuring lengths up to 305 m (1000 feet)
  • Use remote ID allows for a complete wire map of the cable pairs
  • Stores up to 1000 results with descriptive names
Ensure your network is working properly by continuously troubleshooting and optimizing the current infrastructure. The Fluke LinkIQ network testers quickly verify cable performance up to 10 Gbps and help solve network connectivity problems. Testers use frequency-based measurements to accurately determine the distance to fault such as opens, shorts, and unterminated cable up to 305 m (1000 ft) and provide a wiremap of the cable under test. Complete wiremap of the cable pairs which identify miswired and split pairs are possible using the included remote ID.
Obtain detailed information on the nearest connected switch on the LinkIQ tester. Switch port test shows port number, switch name, and port VLAN along with advertised speed and duplex settings. Switch port Power over Ethernet (PoE) setting shows pairs used, power and class available, and results of the PoE test under load. To ensure that the advertised power is actually being delivered by the switch across the cabling infrastructure, the LinkIQ tester will place a load on the connection.
Store up to 1000 results in the tester with descriptive names for documentation. For added convenience, test names and numbers automatically increment as each is saved saving time when testing cables in sequence. Export data to a PC for documentation using LinkWare™ PC cable test management software. LinkWare software saves, maintains, and archives results to create various types of summary reports as well as generate PDFs.
$4,356.45 - $23,538.07CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Battery Range Description
Availability Pricing
Fluke Networks LinkIQ™ Cable + Network Tester 1778469 Mfr # LIQ-100
COMPARE
Li-ion, 3.6 V, 6400 mAh 305 m (1000 ft) LinkIQ™ Cable + Network Tester
$4,356.45CAD / Each
Fluke Networks LinkIQ™ Cable + Network Tester Kit; 1/EA 1778471 Mfr # LIQ-KIT
COMPARE
Li-ion, 3.6 V, 6400 mAh 305 m (1000 ft) LinkIQ™ Cable + Network Tester Kit; 1/EA
$5,231.34CAD / Each
Fluke Networks LinkIQ™ Industrial Ethernet Cable + Network Tester 1778472 Mfr # LIQ-100-IE
COMPARE
Li-ion, 3.6 V, 6400 mAh 305 m (1000 ft) LinkIQ™ Industrial Ethernet Cable + Network Tester
$5,888.77CAD / Each
Fluke Networks LinkIQ™ Industrial Ethernet Cable + Network Tester Kit 1778473 Mfr # LIQ-KIT-IE
COMPARE
Li-ion, 3.6 V, 6400 mAh 305 m (1000 ft) LinkIQ™ Industrial Ethernet Cable + Network Tester Kit
$6,760.65CAD / Each
Fluke Networks LinkIQ™ Kit & Fiber Verification Kit with Inspection (MM & SM) 1778479 Mfr # LIQ-KIT-FTK1475
COMPARE
Li-ion, 3.6 V, 6400 mAh 305 m (1000 ft) LinkIQ™ Kit & Fiber Verification Kit with Inspection (MM & SM)
$17,764.29CAD / Each
Fluke Networks LinkIQ™ Kit with MM Fiber Fault location, Inspection, & Cleaning 1778480 Mfr # LIQ-KIT-FQM-100MIC
COMPARE
Li-ion, 3.6 V, 6400 mAh 305 m (1000 ft) LinkIQ™ Kit with MM Fiber Fault location, Inspection, & Cleaning
$12,313.78CAD / Each
Fluke Networks LinkIQ™ Copper Fiber Kit with Fiber VFL & Live Fiber Detector 1778481 Mfr # LIQ-KIT-VFL-FBRLRT
COMPARE
Li-ion, 3.6 V, 6400 mAh 305 m (1000 ft) LinkIQ™ Copper Fiber Kit with Fiber VFL & Live Fiber Detector
$5,361.59CAD / Each
Fluke Networks LinkIQ™ Twisted Pair & Coax Network Kit with MicroScanner™ 2 Cable Verifier 1778482 Mfr # LIQ-KIT-MS2-100
COMPARE
Li-ion, 3.6 V, 6400 mAh 305 m (1000 ft) LinkIQ™ Twisted Pair & Coax Network Kit with MicroScanner™ 2 Cable Verifier
$5,444.40CAD / Each
Fluke Networks LInkIQ™ Technicians Kit with IS60 Pro-Tool Kit 1778483 Mfr # LIQ-KIT-IS60
COMPARE
Li-ion, 3.6 V, 6400 mAh 305 m (1000 ft) LInkIQ™ Technicians Kit with IS60 Pro-Tool Kit
$5,399.90CAD / Each
Fluke Networks LinkIQ™ Cable + Network Tester Kits; 3/PK 1778485 Mfr # LIQ-KIT-3 Pack
COMPARE
Li-ion, 3.6 V, 6400 mAh 305 m (1000 ft) LinkIQ™ Cable + Network Tester Kits; 3/PK
$13,337.35CAD / Each
Fluke Networks LinkIQ™ Cable + Network Tester Kits; 6/PK 1778486 Mfr # LIQ-KIT-6 Pack
COMPARE
Li-ion, 3.6 V, 6400 mAh 305 m (1000 ft) LinkIQ™ Cable + Network Tester Kits; 6/PK
$23,538.07CAD / Each
Showing 1- 11 of 11