Fluke 566, 568,568ex and 572 Infrared Thermometers
Ideal for maintenance diagnostics in demanding environments
  • Laser targets your measurement area
  • For special surfaces such as concrete all Fluke models have adjustable emissivity (except Fluke 63)
These infrared thermometers are extremely easy to use: just aim, pull the trigger, and read the display. The backlit display shows temperature readings in °F or °C; low-battery indication; and hold, scan, and max. Display holds for seven seconds. Under range and over-range conditions are also indicated on the display.
Available for sale in the United States and Canada only.
$854.55 - $3,860.27CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Emissivity Max Temperature (° F)
Availability Pricing
Fluke 566 Infrared Thermometer, Advanced Model 3563951 Mfr # FLUKE-566
COMPARE
30:1 Adjustable 1202
$854.55CAD / Each
Fluke 568 Infrared Thermometer, Advanced Model 3563961 Mfr # FLUKE-568
COMPARE
50:1 Adjustable 1472
$913.49CAD / Each
Fluke 568Ex Intrinsically Safe Infrared Thermometer, Advanced Model 3563962 Mfr # Fluke 568 EX/ETL
COMPARE
50:1 Adjustable 1472
$3,860.27CAD / Each
Fluke 572-2 Infrared Thermometer, Advanced High-Temperature Model 3563971 Mfr # FLUKE-572-2
COMPARE
60:1 Adjustable 1652