Fluke 568Ex Intrinsically Safe Infrared Thermometer, Advanced Model
Representative image only.
Fluke 568Ex Intrinsically Safe Infrared Thermometer, Advanced Model
Fluke | Mfr: Fluke 568 EX/ETL| Item: 3563962
Ideal for maintenance diagnostics in demanding environments
$3,860.27CAD / Each
for an additional
$310.52CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$3,860.27CAD / Each
Buy It Together
Current Item
Fluke 568Ex Intrinsically Safe Infrared Thermometer, Advanced Model
Fluke 568Ex Intrinsically Safe Infrared Thermometer, Advanced Model
3563962
$3,860.27CAD / Each
+ + =
Total Price 4307.82 ADD 3 ITEMS TO CART
Product Description
Includes
one 9 V battery, CD-ROM user's guide, and hard carrying case. The Fluke 566 adds a type K beaded thermocouple probe, with the Fluke 568 adding s USB cable and FlukeView software.
Key Features
  • Laser targets your measurement area
  • For special surfaces such as concrete all Fluke models have adjustable emissivity (except Fluke 63)
  • 2 year warranty
More About this Item
These infrared thermometers are extremely easy to use: just aim, pull the trigger, and read the display. The backlit display shows temperature readings in °F or °C; low-battery indication; and hold, scan, and max. Display holds for seven seconds. Under range and over-range conditions are also indicated on the display.
Available for sale in the United States and Canada only.
Fluke models 566 and 568 offer additional features including an innovative graphical display, adjustable emissivity through an easily selectable built-in material table , type K thermocouple input, datalogging, multiple language interface, two backlighting levels, and a rubberized housing that improve resistance to shock, damage, water, and dust. These meters also indicate min/max, dif, avg, two color flashing with audio high and low alarms, probe, log, and recall. The 568Ex is Intrinsically safe and has built in data logging up to 99 data points.
Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.
However, none of our infrared thermometers, no matter the manufacture, are approved for medical usage, and are not FDA approved.
For help in selecting an infrared thermometer for industrial, food or laboratory usage please contact the customer service team at Cole-Parmer.
Two year warrantyETL ListedIntrinsically SafeLaser Class II
Specifications
  • Distance To Target Size Ratio (D:S)50:1
  • EmissivityAdjustable
  • Sighting TypeSingle-point laser
  • Laser ClassClass II
  • Min Temperature (° F)-25
  • Max Temperature (° F)1472
  • Min Temperature (° C)-32
  • Max Temperature (° C)800
  • Accuracy±1% or reading or ±2.0°F (±1.0°C), whichever is greater; ±2.0°F (±1.0°C) above 32°F (0°C)
  • Resolution0.1°
  • Response time<500 milliseconds
  • Probe TypeType-K
  • Probe Min Temperature (° F)-454
  • Probe Max Temperature (° F)2501
  • Probe Min Temperature (° C)-270
  • Probe Max Temperature (° C)1372
  • Display TypeBacklit LCD
  • Low Battery IndicatorYes
  • BatteryAA Battery (2)
  • Temperature InputYes
  • DescriptionIntrinsically Safe Infrared Thermometer, Advanced Model