Fluke 572-2 Infrared Thermometer, Advanced High-Temperature Model
Representative image only.
Fluke 572-2 Infrared Thermometer, Advanced High-Temperature Model
Fluke | Mfr: FLUKE-572-2| Item: 3563971
Ideal for maintenance diagnostics in demanding environments
Call For Price
for an additional
$310.52CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Fluke 572-2 Infrared Thermometer, Advanced High-Temperature Model
Fluke 572-2 Infrared Thermometer, Advanced High-Temperature Model
3563971
Call For Price
+ + =
Total Price 447.55 ADD 3 ITEMS TO CART
Product Description
Includes
one 9 V battery, CD-ROM user's guide, and hard carrying case. The Fluke 566 adds a type K beaded thermocouple probe, with the Fluke 568 adding s USB cable and FlukeView software.
Key Features
  • Laser targets your measurement area
  • For special surfaces such as concrete all Fluke models have adjustable emissivity (except Fluke 63)
  • 2 year warranty
More About this Item
These infrared thermometers are extremely easy to use: just aim, pull the trigger, and read the display. The backlit display shows temperature readings in °F or °C; low-battery indication; and hold, scan, and max. Display holds for seven seconds. Under range and over-range conditions are also indicated on the display.
Available for sale in the United States and Canada only.
Fluke model 572-2 offer additional features including an innovative graphical display, adjustable emissivity through an easily selectable built-in material table , type K thermocouple input, datalogging, multiple language interface, two backlighting levels, and a rubberized housing that improve resistance to shock, damage, water, and dust. These meters also indicate min/max, dif, avg, two color flashing with audio high and low alarms, probe, log, and recall. The 572-2 has expanded datalogging capabilities, with internal memory for up to 99 datapoints, or unlimited datapoints through the USB interface direct to your PC. Easy trend analyze can be done through the use of FlukeView software. It also has an expanded temperature range of -22 to 1652°F (-30 to 900°c) and a distance to sight ratio of 60:1.
Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.
However, none of our infrared thermometers, no matter the manufacture, are approved for medical usage, and are not FDA approved.
For help in selecting an infrared thermometer for industrial, food or laboratory usage please contact the customer service team at Cole-Parmer.
Two year warranty
Specifications
  • Distance To Target Size Ratio (D:S)60:1
  • EmissivityAdjustable
  • Sighting TypeDual-point lasers
  • Laser ClassClass II
  • Min Temperature (° F)-22
  • Max Temperature (° F)1652
  • Min Temperature (° C)-30
  • Max Temperature (° C)900
  • Response time<500 milliseconds
  • Display TypeBacklit LCD
  • Battery1 9V (included)
  • DescriptionInfrared Thermometer, Advanced High-Temperature Model