Extech EA30 EasyView Wide-Range Light Meter
Measures up to 400,000 Lux
This meter measures light intensity up to 400,000 lux in five ranges and 40,000 footcandles in four ranges. Unit is color and cosine corrected. Large LCD simultaneously displays readings numerically and in a bar graph format. Meter utilizes a precision silicon photodiode sensor to obtain readings.
$352.14 - $639.45CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Certifications Function Max Measurement (Lux)
Availability Pricing
Extech EA30 Easyview Wide-Range Light Meter 0158865 Mfr # EA30
COMPARE
None Data hold, min/max, peak mode, relative zero function, over-range indicator, auto power-off, low battery indicator 400000
$352.14CAD / Each
Extech EA30-NIST Easyview Wide-Range Light Meter with NIST-Traceable Certificate 0158866 Mfr # EA30-NIST
COMPARE
NIST-Traceable Data hold, min/max, peak mode, relative zero function, over-range indicator, auto power-off, low battery indicator 400000
$639.45CAD / Each