Disposable Bio-Polyethylene Food Grade Sampling Spoons
Ecologically sustainable labware constructed of renewable raw materials
  • Packaged individually and suitable for food-grade applications
  • Produced and packaged according to ISO Class 7
  • EU foodstuffs and FDA compliant
These disposable sampling spoons are ideal for sampling powders, granulates, or liquids. Available nonsterilized or presterilized via gamma irradiation.
$140.41 - $149.55CAD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (in) Sterile
Availability Pricing
Burkle 5379-1011 Disposable sampling spoon, PE, FDA compliant, white, sterile; 2.5 mL 0604294 Mfr # 5379-1011
COMPARE
2.5 5 Yes
$140.41CAD / Pkg of 100
Burkle 5379-1012 Disposable sampling spoon, PE, FDA compliant, white, sterile; 10 mL 0604295 Mfr # 5379-1012
COMPARE
10 6 45/64 Yes
$149.55CAD / Pkg of 100