Digi-Sense Thermocouple Bendable Industrial Probes - MgO Insulated
Davis Instruments can provide many additional configurations beyond those listed here
  • 316 stainless steel sheath
Magnesium Oxide insulated thermocouples, commonly referred to as MgO thermocouples, are used in many process and laboratory applications. Many desirable characteristics make them an excellent choice for general and special purpose applications. They are available in all thermocouple element types, single or dual elements, a wide variety of sheath diameters / materials, rugged in nature, bendable, and have fairly high temperature ratings. The thermocouples listed are commonly used; however, Davis Inotek Instruments can provide many additional configurations beyond those listed.
$102.55 - $170.18CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Sheath Material
Availability Pricing
COMPARE
$170.18CAD / Each
COMPARE
$125.68CAD / Each
COMPARE
$102.55CAD / Each
COMPARE
$125.68CAD / Each
COMPARE
$102.55CAD / Each