CYTIVA Whatman™ Air Sampling Filters
Meet requirements for EPA PM-10 micron and PM-2.5 micron testing
  • Ideal for air particle monitoring and environmental testing
Air Sampling Filters for high-volume EPA PM-10 sampling equipment are used to collect atmospheric particulates and aerosols. Pure borosilicate glass fiber allows detailed chemical analysis of trace pollutants with minimum interference or background.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
$140.49 - $1,628.10CAD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Material Pore Size (µm) Description
Availability Pricing
Whatman 1882-047 Air Sampling Filters, EPM, 4.7 cm Dia 2970600 Mfr # 1882-047
COMPARE
Borosilicate Glass 2.5 Air Sampling Filters, EPM, 4.7 cm Dia
$140.49CAD / Pkg of 100
Whatman 1882-866 Air Sampling Filters, EPM, 8"W x 10"H 2970606 Mfr # 1882-866
COMPARE
Borosilicate Glass 2.5 Air Sampling Filters, EPM, 8"W x 10"H
$1,628.10CAD / Pkg of 100