Whatman 1882-866 Air Sampling Filters, EPM, 8"W x 10"H
Representative image only.
Whatman 1882-866 Air Sampling Filters, EPM, 8"W x 10"H
Whatman | Mfr: 1882-866| Item: 2970606
Meet requirements for EPA PM-10 micron and PM-2.5 micron testing
$1,628.10CAD / Pkg of 100
$1,628.10CAD / Pkg of 100
Product Description
Key Features
  • Ideal for air particle monitoring and environmental testing
More About this Item
Air Sampling Filters for high-volume EPA PM-10 sampling equipment are used to collect atmospheric particulates and aerosols. Pure borosilicate glass fiber allows detailed chemical analysis of trace pollutants with minimum interference or background.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
Specifications
  • Membrane MaterialBorosilicate Glass
  • Pore Size (µm)2.5
  • DescriptionAir Sampling Filters, EPM, 8"W x 10"H