Corning Digital Ceramic Top Hot Plates
Feature international symbols for easy recognition by all operators
  • Microprocessor-controlled feedback maintains constant temperature
  • Small footprint and low profile conserves lab space and easily fits inside a hood or cabinet
Flat, bright white Pyroceram® brand glass ceramic top material provides uniform heating, is easy to clean, and resists scratches, corrosion, and chemicals. The housing features a spill trough to deflect spills away from the display and control knobs. Electronics are sealed and enclosed to protect against harsh chemicals. An external support rod holder let you mount a thermometer, electrode, or burette. Hot plates feature a LED light to warn you if temperature exceeds 140°F (60°C). Temperature display is adjustable in 5° increments and indicates when set temperature is reached.
$857.51 - $1,249.43CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Length (in)
Availability Pricing
Corning 6798-400D Digital Hot Plate, 5" x 7", 220 VAC 8430135 Mfr # 6798-400D
COMPARE
220 5
$857.51CAD / Each
Corning 6795-600D Digital Hot Plate, 10" x 10", 120 VAC 8430160 Mfr # 6795-600D
COMPARE
120 10
$1,249.43CAD / Each
Corning 6798-600D Digital Hot Plate, 10" x 10", 220 VAC 8430165 Mfr # 6798-600D
COMPARE
220 10
$1,043.16CAD / Each