Corning Calibrated Graduated Cylinders, PP
  • Meet ISO Standard 6706
Manufactured of translucent polpypropylene, these sturdy cylinders feature raised graduations and a wide hexagonal base. Calibrated "to contain" at 20°C. Autoclavable; exposure to temperatures above 140°F (60°C) may affect accuracy.
$324.15 - $552.52CAD / Case of 6
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Corning 3022P-250 Graduated Cylinder 250 mL PP, 6/pk 0613739 Mfr # 3022P-250
COMPARE
250 2 Polypropylene
$324.15CAD / Case of 6
Corning 3022P-500 Graduated Cylinder 500 mL PP, 6/pk 0613741 Mfr # 3022P-500
COMPARE
500 5 Polypropylene
$421.39CAD / Case of 6
Corning 3022P-1L Calibrated 'To Contain' PP Graduated Cylinders, 1 L, 6/Pk 0613743 Mfr # 3022P-1L
COMPARE
1000 10 Polypropylene
$552.52CAD / Case of 6
Corning 3022P-2L Calibrated 'To Contain' PP Graduated Cylinders, 2 L, 3/Pk 0613745 Mfr # 3022P-2L
COMPARE
2000 20 Polypropylene
$468.54CAD / Case of 3