Corning 3022P-2L Calibrated 'To Contain' PP Graduated Cylinders, 2 L, 3/Pk

Representative image only.

Corning 3022P-2L Calibrated 'To Contain' PP Graduated Cylinders, 2 L, 3/Pk

Corning | Mfr: 3022P-2L| Item: 0613745

$468.54CAD / Case of 3

$468.54CAD / Case of 3
Product Description

Key Features

  • Meet ISO Standard 6706

More About this Item

Manufactured of translucent polpypropylene, these sturdy cylinders feature raised graduations and a wide hexagonal base. Calibrated "to contain" at 20°C. Autoclavable; exposure to temperatures above 140°F (60°C) may affect accuracy.

Specifications
  • MaterialPolypropylene
  • Capacity (mL)2000
  • Graduation Interval (mL)20
  • AutoclavableYes
  • DescriptionCalibrated 'To Contain' PP Graduated Cylinders, 2 L, 3/Pk
Reviews
Q&A