Cole-Parmer® VapLock™ Solvent Waste Kits with Carboy
Complete solution to safely collect effluent from multiple sources while minimizing harmful solvent exhaust
  • 10 port manifold—1 x 1/2" NPT(F), 3 x 1/4" NPT(F), and 6 x 1/4-28 UNF(F)
  • Designed to provide an effective means of sealed solvent delivery
  • For collection of solvent waste from multiple effluent streams
The Cole-Parmer® VapLock™ solvent waste kits with carboy include all the required components for safe waste containment. Use the waste kits to minimize leaks, spills, and vapor escape from solvent waste containers. Waste kits include a 10-port manifold, exhaust filter, adapters, plugs, tubing, and container—everything you need to install on a typical HPLC instrument.
Manifold allows solvent waste to be collected from multiple effluent streams. Exhaust filter minimizes exposure to harmful chemical vapors. Fittings and tubing provide leak-tight connections minimizing spills and leaks. Durable plastic containers allow you to meet your large volume liquid storage needs. Integrated wide handles make them easy to carry and pour, even with gloves on.
$575.36 - $780.90CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Special Features
Availability Pricing
COMPARE
10 100-415 Round Wide-Mouth Carboy
$698.39CAD / Each
COMPARE
20 100-415 Round Wide-Mouth Carboy
$780.90CAD / Each
COMPARE
10 83B Round Carboy
$698.39CAD / Each
COMPARE
20 83B Round Carboy
$739.64CAD / Each
COMPARE
10 63 mm Square Carboy
$575.36CAD / Each
COMPARE
10 53B Jerrican
$723.43CAD / Each
COMPARE
20 53B Jerrican
$739.64CAD / Each
COMPARE
5 GL45 Heavy-Duty Square Carboy
$657.13CAD / Each
Showing 1- 8 of 8