Cole-Parmer® Split Tube Furnaces
Split hinged body design provides easy process tube installation and changes
  • Vacuum formed ceramic fiber heating elements offer excellent temperature uniformity
  • Set up in horizontal or vertical orientation
  • Single-zone configuration
  • Long life Inconel sheathed type K thermocouple
Split tube furnaces feature a body that is split in half, attached by a hinge, allowing easy process tube installation and removal; use process tubes with end seals attached. Set up tube furnaces for use in either horizontal or vertical orientation. The vacuum-formed ceramic fiber heating elements with free radiating wire embedded into the body of the heater to ensure rapid heat up, excellent temperature uniformity, and quick cooldown. Select from compact and lightweight split tube furnace with 1100 °C (2012 °F) maximum operating temperature or larger split tube furnaces with 1200°C (2192 °F) maximum operating temperature.
$5,201.07 - $17,282.86CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Process Tube OD (in)
Availability Pricing
COMPARE
1200 120 1.25
$5,201.07CAD / Each
COMPARE
1200 208 / 240 1.25
$5,245.27CAD / Each
COMPARE
1200 230 3
$9,157.12CAD / Each
COMPARE
1200 230 3
$12,855.33CAD / Each
COMPARE
1200 230 6
$17,282.86CAD / Each