Cole-Parmer® Nylon (Polyamide) Membranes
Ideal for filtering or sterilizing nonprotein-containing biological solutions
  • High protein binding
  • Withstand repeated steam sterilization
Inherently hydrophilic membranes do not require prewetting — use to filter aqueous solutions and organic solvents.
$83.25 - $401.50CAD / Pkg of 100
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Pore Size (µm)
Availability Pricing
COMPARE
13 0.2
$188.96CAD / Pkg of 100
COMPARE
47 0.2
$320.46CAD / Pkg of 100
COMPARE
90 0.2
$219.54CAD / Pkg of 25
COMPARE
142 0.2
$388.24CAD / Pkg of 25
COMPARE
13 0.45
$171.65CAD / Pkg of 100
COMPARE
47 0.45
$307.94CAD / Pkg of 100
COMPARE
90 0.45
$168.33CAD / Pkg of 25
COMPARE
13 0.2
$83.25CAD / Pkg of 100
COMPARE
25 0.2
$196.99CAD / Pkg of 100
COMPARE
47 0.2
$294.46CAD / Pkg of 100
COMPARE
90 0.2
$330.33CAD / Pkg of 25
COMPARE
25 0.45
$196.99CAD / Pkg of 100
COMPARE
47 0.45
$401.50CAD / Pkg of 100
COMPARE
90 0.45
$133.86CAD / Pkg of 25
Showing 1- 14 of 14