Cole-Parmer® Nitrocellulose (NC) Filters
Nitrocellulose (NC) filters exhibit the highest protein retention of all membranes—good binding matrix for macromolecules
  • Provide fast flow rates with aqueous solutions
Filters are Triton-free and have a bidirectional membrane support and luer lock inlet/outlet for a secure fit to the syringe. Membrane type and pore size are printed on each filter for easy identification. All Cole-Parmer syringe filters are 100% integrity tested and manufactured in accordance with ISO 9002 standards. Sterile filters are sterilized by gamma radiation. They are certified nonpyrogenic, noncytotoxic, and are blister packed.
$279.72 - $603.35CAD / Box of 50
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
COMPARE
25 0.22 Yes
$279.72CAD / Box of 50
COMPARE
25 0.45 Yes
$297.04CAD / Pkg of 50
COMPARE
25 0.2 No
$603.35CAD / Box of 100