Cole-Parmer® Gel-Loading Tips
Get easy loading of electrophoresis gels
  • Easily fit into small instrument openings
  • Ideal for DNA sequencing
These extended microcapillary tips are excellent for loading electrophoresis gels. The polypropylene tips are rounded to fit easily into small instrument openings. Use these microcapillary tips for DNA sequencing, loading SDS PAGE gels, and other critical sampling applications.
$159.13 - $266.68CAD / Pkg of 1020
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Package Of
Availability Pricing
COMPARE
200 1 5 Racks; 204/pk
$266.68CAD / Pkg of 1020
COMPARE
10 0.1 Bulk; 1000/pk
$159.13CAD / Pkg of 1000
COMPARE
10 0.1 5 Racks; 204/pk
$262.26CAD / Pkg of 1020