CAPP® CappOrigami™ Multichannel Pipette Reservoirs
Innovative design minimizes reagent waste and saves valuable storage space
  • Designed to be shipped and stored as flat sheets to save space and reduce shipping costs
  • Constructed of durable and chemical-resistant polypropylene
CAPP® CappOrigami™ reagent reservoirs are designed to be shipped and stored as flat sheets to save space and reduce shipping costs. The polypropylene construction features a sharp V-shape that makes pipetting easier, prevents the formation of bubbles in the pipette tips, and allows you to use more accurate amounts of reagent and avoid expensive waste. The easy- to-use design features prefolded lines — simply push the sides to ''assemble''. The folds ensure that the reservoirs stay upright.
$135.99 - $177.18CAD / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compatible with Max Volume (mL) Sterile
Availability Pricing
Capp CA40506 Reagent Reservoir sterile 30 mL,10 x 5/bag 2501962 Mfr # CA40506
COMPARE
8- and 16-channel pipettors 30 Yes
$155.44CAD / Pkg of 10
CappOrigami™ Reagent Reservoir, Polypropylene, Non-Sterile, 40 mL; 50/Pk 2501964 Mfr # CA40510
COMPARE
12-channel pipettors 40 No
$135.99CAD / Pkg of 50
CappOrigami™ Reagent Reservoir, Polypropylene, Sterile, 40 mL; 5/Bag, 10 Bags/Pk 2501966 Mfr # CA40511
COMPARE
12-channel pipettors 40 Yes
$177.18CAD / Pkg of 50