CappOrigami™ Reagent Reservoir, Polypropylene, Non-Sterile, 40 mL; 50/Pk
Representative image only.
CappOrigami™ Reagent Reservoir, Polypropylene, Non-Sterile, 40 mL; 50/Pk
Capp | Mfr: CA40510| Item: 2501964
Innovative design minimizes reagent waste and saves valuable storage space
$135.99CAD / Pkg of 50
$135.99CAD / Pkg of 50
Product Description
Key Features
  • Designed to be shipped and stored as flat sheets to save space and reduce shipping costs
  • Constructed of durable and chemical-resistant polypropylene
  • Accommodates 12-channel pipettors
  • 3 year warranty
More About this Item
CAPP® CappOrigami™ reagent reservoirs are designed to be shipped and stored as flat sheets to save space and reduce shipping costs. The polypropylene construction features a sharp V-shape that makes pipetting easier, prevents the formation of bubbles in the pipette tips, and allows you to use more accurate amounts of reagent and avoid expensive waste. The easy- to-use design features prefolded lines — simply push the sides to ''assemble''. The folds ensure that the reservoirs stay upright.
3 Year Warranty
Specifications
  • SterileNo
  • Max Volume (mL)40
  • Wetted MatterialPolypropylene
  • Package Of50/Pk
  • Compatible with12-channel pipettors
  • DescriptionReagent Reservoir, Polypropylene, Non-Sterile, 40 mL; 50/Pk