Capp CA40506 Reagent Reservoir sterile 30 mL,10 x 5/bag

Representative image only.

Capp CA40506 Reagent Reservoir sterile 30 mL,10 x 5/bag

Capp | Mfr: CA40506| Item: 2501962

Innovative design minimizes reagent waste and saves valuable storage space

$155.44CAD / Pkg of 10

$155.44CAD / Pkg of 10
Product Description

Key Features

  • Designed to be shipped and stored as flat sheets to save space and reduce shipping costs
  • Constructed of durable and chemical-resistant polypropylene
  • 3 year warranty

More About this Item

CAPP® CappOrigami™ reagent reservoirs are designed to be shipped and stored as flat sheets to save space and reduce shipping costs. The polypropylene construction features a sharp V-shape that makes pipetting easier, prevents the formation of bubbles in the pipette tips, and allows you to use more accurate amounts of reagent and avoid expensive waste. The easy- to-use design features prefolded lines — simply push the sides to ''assemble''. The folds ensure that the reservoirs stay upright.

3 Year Warranty
Specifications
  • SterileYes
  • Max Volume (µL)30000
  • Min Volume (µL)0
  • Min Volume (mL)0
  • Max Volume (mL)30
  • Compatible with8- and 16-channel pipettors
  • DescriptionReagentReservoirsterile30mL10x5/bag
  • Warranty3 year
Reviews
Q&A