BriskHeat® SpeedTrace Extreme Pre-Assembled Self-Regulating Heating Cables
Ideal for freeze protection and thawing: metal or plastic pipes and valves
  • Suitable for temperatures down to: -40°F (-40°C)
  • Easy-to-install: pre-assembled with power cord and plug
  • Automatically adjusts heat output based on surface and ambient temperature
  • No temperature controller is required
  • Safe to overlap the cable and insulate
$82.07 - $402.24CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat FFSL81-6 Speedtrace Extreme Heating Cable, 8 Watts/ft, 120V, 6ft with Plug 3611540 Mfr # FFSL81-6
COMPARE
150 120
$82.07CAD / Each
BriskHeat FFSL81-12 Speedtrace Extreme Heating Cable, 8 Watts/ft, 120V, 12ft with Plug 3611541 Mfr # FFSL81-12
COMPARE
150 120
$98.72CAD / Each
BriskHeat FFSL81-18 Speedtrace Extreme Heating Cable, 8 Watts/ft, 120V, 18ft with Plug 3611542 Mfr # FFSL81-18
COMPARE
150 120
$116.84CAD / Each
BriskHeat FFSL81-24 Speedtrace Extreme Heating Cable, 8 Watts/ft, 120V, 24ft with Plug 3611543 Mfr # FFSL81-24
COMPARE
150 120
$137.47CAD / Each
BriskHeat FFSL81-50 Speedtrace Extreme Heating Cable, 8 Watts/ft, 120V, 50ft with Plug 3611544 Mfr # FFSL81-50
COMPARE
150 120
$227.64CAD / Each
BriskHeat FFSL81-75 Speedtrace Extreme Heating Cable, 8 Watts/ft, 120V, 75ft with Plug 3611545 Mfr # FFSL81-75
COMPARE
150 120
$317.15CAD / Each
BriskHeat FFSL81-100 Speedtrace Extreme Heating Cable, 8 Watts/ft, 120V, 100ft with Plug 3611546 Mfr # FFSL81-100
COMPARE
150 120
$402.24CAD / Each
Showing 1- 7 of 7