Boekel Electronic Desiccators with Stainless Steel Shelves
Don’t keep replacing desiccant
  • Automatic regeneration of desiccant every five hours
No wasted space—desiccant housed in side mount box for more sample space
– Sturdy, stainless steel and glass construction
– Monitor humidity with included hygrometer
$5,532.58 - $8,744.57CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Exterior Height (InchShort) Exterior Width (in) Power (VAC)
Availability Pricing
Boekel Scientific 134241-2 Electronic Desiccator with SS Shelves; 11.5" x 12" x 10"D; 230V 0890135 Mfr # 134241-2
COMPARE
12.25 16 230
$5,532.58CAD / Each
Boekel Scientific 134441-2 Electronic Desiccator with SS Shelves, 230 VAC 0890145 Mfr # 134441-2
COMPARE
22.25 20.5 230
$8,744.57CAD / Each