Binder BF Series Mechanical Convection Incubators
Exceptional efficiency and perfect temperature accuracy thanks to the very latest APT.line™ technology
  • Class 3.1 independent temperature safety device (DIN 12880) with visual temperature alarm
  • Adjustable exhaust air flap
  • Inner tempered glass door for quick viewing of samples
These mechanical convection incubators are suitable for all gentle incubation applications, particularly under a full load and large batch throughputs. The intuitive controller with integrated timer function makes it easy to control operating parameters including exhaust flap and fan speed. The adjustable fan moves air evenly and efficiently providing very little temperature variation throughout the unit.
$4,535.09 - $9,216.05CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Binder BF56 Avantgarde Mechanical Convection Incubator, 2.1 cu ft, 120 VAC 3907303 Mfr # 9010-0314
COMPARE
2.1 120
$4,535.09CAD / Each
Binder BF115 Avantgarde Mechanical Convection Incubator, 4 cu ft, 120 VAC 3907307 Mfr # 9010-0316
COMPARE
4 120
$6,214.76CAD / Each
Binder BF260 Avantgarde Mechanical Convection Incubator, 9.1 cu ft, 120 VAC 3907311 Mfr # 9010-0320
COMPARE
9.1 120
$9,216.05CAD / Each
Binder BF400 Classic Mechanical Convection Incubator, 14.3 cu ft, 115 VAC 3907316 Mfr # 9010-0242
COMPARE
14.3 120