Ansell Conform Latex Gloves
Reliable barrier protection with maximum degree of dexterity
  • Tear resistance
  • Ideal for general laboratory
Natural rubber latex gloves are 3.5-mil thick for strength and tear-resistance. Inner polymer coating for easy donning and doffing. General purpose gloves are ideal for general laboratory, parts handling, electronics assembly, or quality control applications. Sold in box of 100.
$28.29 - $30.79CAD / Box of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
Ansell TouchNTuff® 69-210 Latex Gloves, Small, 100/bx 8656800 Mfr # 69-210-516704
COMPARE
9 1/2 Small
$30.79CAD / Box of 100
Ansell TouchNTuff® 69-210 Latex Gloves, Medium, 100/bx 8656802 Mfr # 516705
COMPARE
9 1/2 Medium
$29.47CAD / Box of 100
Ansell TouchNTuff® 69-210 Latex Gloves, Large, 100/bx 8656804 Mfr # 69-210-516706
COMPARE
9 1/2 Large
$28.29CAD / Box of 100