Ansell TouchNTuff® 69-210 Latex Gloves, Medium, 100/bx
Representative image only.
Ansell TouchNTuff® 69-210 Latex Gloves, Medium, 100/bx
Ansell | Mfr: 516705| Item: 8656802
Reliable barrier protection with maximum degree of dexterity
$29.47CAD / Box of 100
BUY MORE & SAVE
  • 1-9$29.47 CAD / Box of 100
  • 10+$26.52 CAD / Box of 100
$29.47CAD / Box of 100
BUY MORE & SAVE
  • 1-9$29.47 CAD / Box of 100
  • 10+$26.52 CAD / Box of 100
Product Description
Key Features
  • Tear resistance
  • Ideal for general laboratory
More About this Item
Natural rubber latex gloves are 3.5-mil thick for strength and tear-resistance. Inner polymer coating for easy donning and doffing. General purpose gloves are ideal for general laboratory, parts handling, electronics assembly, or quality control applications. Sold in box of 100.
Specifications
  • Thickness (mil)5
  • MaterialLatex
  • SizeMedium
  • Length (in)9 1/2
  • Length (cm)24
  • ColorWhite
  • CuffBeaded
  • SterileNo
  • DescriptionLatex Gloves, medium, 100/bx