Ansell TouchNTuff® 69-210 Latex Gloves, Large, 100/bx
Representative image only.
Ansell TouchNTuff® 69-210 Latex Gloves, Large, 100/bx
Ansell | Mfr: 69-210-516706| Item: 8656804
Reliable barrier protection with maximum degree of dexterity
$28.29CAD / Box of 100
BUY MORE & SAVE
  • 1-9$28.29 CAD / Box of 100
  • 10+$25.46 CAD / Box of 100
$28.29CAD / Box of 100
BUY MORE & SAVE
  • 1-9$28.29 CAD / Box of 100
  • 10+$25.46 CAD / Box of 100
Product Description
Key Features
  • Tear resistance
  • Ideal for general laboratory
More About this Item
Natural rubber latex gloves are 3.5-mil thick for strength and tear-resistance. Inner polymer coating for easy donning and doffing. General purpose gloves are ideal for general laboratory, parts handling, electronics assembly, or quality control applications. Sold in box of 100.
Specifications
  • MaterialLatex
  • Thickness (mil)5
  • SizeLarge
  • Length (in)9 1/2
  • Length (cm)24
  • ColorWhite
  • CuffBeaded
  • SterileNo
  • DescriptionLatex Gloves, large, 100/bx