Analtech Glass-Backed Analytical TLC Uniplates
Efficient separations — economically!
  • Constructed of glass backing
These TLC plates combine top-quality adsorbents, consistent manufacturing, rigid QC, and careful packaging--the results of 40 years of experience dedicated to thin layer techniques. Select the perfect analytical plates for your application. Choose plates with or without an acid stable fluorescent indicator (UV 254 nm). Scored plates snap apart evenly to facilitate single-use separations or screening.
$95.03 - $717.54CAD / Pkg of 25
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
Compareitem specifications
Layer Thickness (micron) Plate Length (cm) Plate Width (cm)
Availability Pricing
Analtech Uniplates, 20x20cm/Soft/Silica G/250um Thick/15um Particle; 25/Pk 3410100 Mfr # P01011
COMPARE
250 20 20
$271.10CAD / Pkg of 25
Analtech Uniplates, 20x20cm/UV/Soft/Silica G/250um Thick/15um Particle; 25/Pk 3410105 Mfr # P02011
COMPARE
250 20 20
$271.10CAD / Pkg of 25
Analtech Uniplates, 20x20cm/Scored/Soft/Silica G/250um Thick/15um Particle; 25/Pk 3410110 Mfr # P01511
COMPARE
250 20 20
$346.25CAD / Pkg of 25
Analtech Uniplates, 20x20cm/UV/Scored/Soft/Silica G/250um Thick/15um Part; 25/Pk 3410115 Mfr # P02511
COMPARE
250 20 20
$346.25CAD / Pkg of 25
Analtech Uniplates, 10x20cm/Soft/Silica G/250um Thick/15um Particle; 25/Pk 3410120 Mfr # P01021
COMPARE
250 10 20
$171.80CAD / Pkg of 25
Analtech Uniplates, 10x20cm/UV/Soft/Silica G/250um Thick/15um Particle; 25/Pk 3410125 Mfr # P02021
COMPARE
250 10 20
$171.80CAD / Pkg of 25
Analtech Uniplates, 10x20cm/Scored/Soft/Silica G/250um Thick/15um Particle; 25/Pk 3410130 Mfr # P01521
COMPARE
250 10 20
$235.01CAD / Pkg of 25
Analtech Uniplates, 10x20cm/UV/Scored/Soft/Silica G/250um Thick/15um Part; 25/Pk 3410135 Mfr # P02521
COMPARE
250 10 20
$235.01CAD / Pkg of 25
Analtech Uniplates, 20 x 20 cm, Hard, Silica HL, 250 um Thick, 10 um Particle; 25/Pk 3410140 Mfr # P46011
COMPARE
250 20 20
$271.10CAD / Pkg of 25
Analtech Uniplates, 20x20cm/UV/Hard/Silica HL/250um Thick/10um Particle; 25/Pk 3410145 Mfr # P47011
COMPARE
250 20 20
$271.10CAD / Pkg of 25
Analtech Uniplates, 20x20cm/Scored/Hard/Silica HL/250um Thick/10um Part; 25/Pk 3410150 Mfr # P46511
COMPARE
250 20 20
$346.25CAD / Pkg of 25
Analtech Uniplates, 10 x 20 cm, Hard, Silica HL, 250 um Thick, 10 um Particle; 25/Pk 3410160 Mfr # P46021
COMPARE
250 10 20
$171.80CAD / Pkg of 25
Analtech Uniplates, 10x20cm/Scored/Hard/Silica HL/250um Thick/10um Part; 25/Pk 3410170 Mfr # P46521
COMPARE
250 20 20
$235.01CAD / Pkg of 25
Analtech Uniplates, 10x20cm/UV/Scored/Hard/Silica HL/250um Thick/10um Part; 25/Pk 3410175 Mfr # P47521
COMPARE
250 20 20
$235.01CAD / Pkg of 25
Analtech Uniplates, 5x20cm/Hard/Silica HL/250um Thick/10um Particle; 25/Pk 3410180 Mfr # P46031
COMPARE
250 5 20
$95.03CAD / Pkg of 25
Analtech Uniplates, 5x20cm/UV/Hard/Silica HL/250um Thick/10um Particle; 25/Pk 3410185 Mfr # P47031
COMPARE
250 5 20
$95.03CAD / Pkg of 25
Analtech Uniplates, 20x20cm/UV/Hard/Silica GHL/250um Thick/15um Particle; 25/Pk 3410205 Mfr # P21011
COMPARE
250 20 20
$271.10CAD / Pkg of 25
Analtech Uniplates, 20x20cm/Scored/Hard/Silica GHL/250um Thick/15um Part; 25/Pk 3410210 Mfr # P11511
COMPARE
250 20 20
$346.25CAD / Pkg of 25
Analtech Uniplates, 10x20cm/Hard/Silica GHL/250um Thick/15um Particle; 25/Pk 3410220 Mfr # P11021
COMPARE
250 10 20
$171.80CAD / Pkg of 25
Analtech Uniplates, 10x20cm/UV/Scored/Hard/Silica GHL/250um Thick/15um Part 25/Pk 3410228 Mfr # P21521
COMPARE
250 10 20
$272.58CAD / Pkg of 25
Showing 1 - 20 of 25