Additel Pneumatic Test Pumps
Unique hand-tightened quick disconnects provide easy hand operation
  • Precision screw press technology provides great stability, eliminates leakage
  • Isothermal bellows chamber yields high pressure stability
  • Unique quick-connect fittings tighten easily by hand, no tools or tape needed
The ergonomic pump design and unique hand-tightened quick disconnects provide easy hand operation of these pneumatic test pumps. Isothermal bellows chamber is not affected by environmental temperature changes. A built-in gas-liquid isolator protects pump from moisture and dirt. Easily connect and disconnect to the test pump without the need for PTFE tape or wrenches.
Variety of models are available to meet your application and budget.
$2,537.18 - $9,798.04CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Pressure Resolution Description
Availability Pricing
Additel 901-N Low Pressure Pneumatic Test Pump, 0 to 6psi 1610601 Mfr # ADT901B-N
COMPARE
6 psi (0.4 bar) 0.01 Pa (0.0001 mbar) Low Pressure Pneumatic Test Pump, 0 to 6psi
$2,537.18CAD / Each
Additel 912-N Low Pressure Pneumatic Test Pump, 95% Vacuum to 60psi 1610602 Mfr # ADT912A-N
COMPARE
60 psi (4 bar) 0.01 Pa (0.0001 mbar) Low Pressure Pneumatic Test Pump, 95% Vacuum to 60psi
$3,543.50CAD / Each
Additel 914-N Handheld Pneumatic Test Pump, 95% Vacuum to 375psi 1610603 Mfr # ADT914A-N
COMPARE
375 psi (25 bar) 0.01 Pa (0.0001 mbar) Handheld Pneumatic Test Pump, 95% Vacuum to 375psi
$2,683.04CAD / Each
Additel 916-N Portable Pneumatic Test Pump, 95% Vacuum to 600psi 1610604 Mfr # ADT916A-N
COMPARE
600 psi (40 bar) 0.01 Pa (0.0001 mbar) Portable Pneumatic Test Pump, 95% Vacuum to 600psi
$3,919.22CAD / Each
Additel 917 Pneumatic Pressure Test Pump, -14 psi to 1000 psi 1610742 Mfr # ADT917-N
COMPARE
1000 psi (70 bar) 0.01 Pa (0.0001 mbar) Pneumatic Pressure Test Pump, -14 psi to 1000 psi
$4,991.85CAD / Each
Additel 918 Pneumatic Pressure Test Pump, -14 psi to 1500 psi 1610743 Mfr # ADT918-N
COMPARE
1500 psi (105 bar) 0.01 Pa (0.0001 mbar) Pneumatic Pressure Test Pump, -14 psi to 1500 psi
$6,065.95CAD / Each
Additel 919A High Pressure Pneumatic Test Pump, -14 to 2000 psi 1610744 Mfr # ADT919A-N
COMPARE
3000 psi (210 bar) 0.01 Pa (0.0001 mbar) High Pressure Pneumatic Test Pump, -14 to 2000 psi
$7,923.89CAD / Each
Additel 920-N High Pressure Pneumatic Test Pump, 95% Vacuum to 3000psi 1610784 Mfr # ADT920-N
COMPARE
3000 psi (210 bar) 10 Pa (0.1 mbar) High Pressure Pneumatic Test Pump, 95% Vacuum to 3000psi
$9,798.04CAD / Each
Showing 1- 8 of 8