Adam Equipment Equinox Semi Micro Balances
Remarkable precision for detailed testing
  • 5-year manufacturer’s warranty
  • Large, bright, clear weight display
  • USB and RS-232 interfaces provide fast communication with computer/printer
  • Sealed keypad protects against dirt and spills
  • Multiple weighing modes including checkweighing, counting, percentage weighing and density determination
  • Selectable digital filtering for animal/dynamic weighing
  • Security slot for optional Kensington-type lock and cable to prevent theft
The Equinox line of semi micro analytical balances provide many features and functions—at an affordable price. Choose from a variety of capacities with external caibration (must use calibration mass to calibrate), or internal calibration (may use with or without a calibration mass).
Easily view results on the 5" (125 mm) display. This color touchscreen also makes function selection simple, and menu navigation is intuitive. Order the optional in-use cover to protect the screen, while still allowing operation.
The level indicator and adjustable feet ensures proper set up to obtain precise measurements. Perform checkweighing, parts counting, percent weighing, density determinations, dynamic weighing, formulation dosing, and statistical analysis.
$4,722.21 - $10,438.97CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Description
Availability Pricing
Adam Equipment EAB 124e Equinox Analytical Balance, 120g x 0.0001g, External Calibration 1112503 Mfr # EAB 124e
COMPARE
External calibration 120 Equinox Analytical Balance, 120g x 0.0001g, External Calibration
$4,722.21CAD / Each
Adam Equipment EAB 224e Equinox Analytical Balance, 220g x 0.0001g, External Calibration 1112505 Mfr # EAB 224e
COMPARE
External calibration 220 Equinox Analytical Balance, 220g x 0.0001g, External Calibration
$5,215.80CAD / Each
Adam Equipment EAB 314E Equinox Analytical Balance, 310g x 0.0001g, External Calibration 1112506 Mfr # EAB 314E
COMPARE
External calibration 310 Equinox Analytical Balance, 310g x 0.0001g, External Calibration
$5,753.59CAD / Each
Adam Equipment EAB 124i Equinox Semi Micro Balance, 120g x 0.0001g, Internal Calibration 1112507 Mfr # EAB 124I
COMPARE
Internal 120 Equinox Semi Micro Analytical Balance, 120g x 0.0001g, Internal Calibration
$5,127.40CAD / Each
Adam Equipment EAB 224i Equinox Analytical Balance, 220g x 0.0001g, Internal Calibration 1112508 Mfr # EAB 224i
COMPARE
Internal automatic calibration 220 Equinox Analytical Balance, 220g x 0.0001g, Internal Calibration
$5,628.35CAD / Each
Adam Equipment EAB 314i Equinox Analytical Balance, 310g x 0.0001g, Internal Calibration 1112509 Mfr # EAB 314i
COMPARE
Internal automatic calibration 310 Equinox Analytical Balance, 310g x 0.0001g, Internal Calibration
$6,144.04CAD / Each
Adam Equipment EAB 414i Equinox Analytical Balance, 410g x 0.0001g, Internal Calibration 1112511 Mfr # EAB 414i
COMPARE
Internal automatic calibration 410 Equinox Analytical Balance, 410g x 0.0001g, Internal Calibration
$8,295.19CAD / Each
Adam Equipment EAB 514i Equinox Semi Micro Balance, 510g x 0.0001g, Internal Calibration 1112512 Mfr # EAB 514I
COMPARE
Internal 510 Equinox Semi Micro Analytical Balance, 510g x 0.0001g, Internal Calibration
$10,438.97CAD / Each
Showing 1- 8 of 8