In Stock
Length (in)
Width (in)
Material
Compliance
Controlled Environment
APPLY FILTERS
filter byclear all
54 RESULTS
SORT BY
Showing 1 - 24 of 54
23>
What is a controlled environment?
A controlled environment is a setting where conditions are managed and regulated to ensure consistency and stability. These conditions include temperature, humidity, light, air quality, and other environmental factors. Controlled environments are crucial in various fields, such as scientific research, manufacturing, and agriculture. For example, laboratories use controlled environments to conduct experiments under stable conditions, and greenhouses maintain specific climates to optimize plant growth. This approach ensures that external variables do not influence the results of an experiment or production process.
What is the difference between a controlled environment and a cleanroom?
All cleanrooms are controlled environments, but not all controlled environments are cleanrooms.
Controlled Environment
A controlled environment is a broad term that refers to any space where specific aspects of the environment (such as temperature, humidity, light, and air quality) are regulated to maintain consistency and stability. These are used in various fields like agriculture, scientific research, and manufacturing to ensure that external factors do not adversely affect processes or experiments. The level of control can vary based on the requirements of the task or experiment being conducted.
Cleanroom
A cleanroom is a type of controlled environment with an extremely high level of contamination control. The primary focus is on minimizing the presence of airborne particles, microbes, and chemical vapors. Cleanrooms are classified by the number of particles allowed per cubic meter, at a specified particle size. They are essential in industries where small particles, such as pharmaceuticals, biotech, and surgical environments, can adversely affect manufacturing.
Controlled Environment
Step into a world where precision meets cleanliness. Our controlled environment solutions are designed for the meticulous demands of life sciences. Discover our extensive collection, including gloves engineered for sensitivity and safety, mats that trap contaminants, HEPA dust bags ensuring air purity, mops that sanitize without a trace, wipes for ultra-clean surfaces, cleanroom swabs for detailed application, and more. Equip your space with our trusted products to maintain an impeccable, controlled environment.
Enhance Safety and Reliability:
Designed with your safety in mind, meeting stringent standards to reduce the risk of contamination.
They ensure reliable results in your sensitive experiments and manufacturing processes, helping you maintain the highest precision and quality.
Comply with Industry Standards:
Crafted to help you meet the regulatory requirements of pharmaceuticals, biotechnology, and manufacturing industries.
Compliance with ISO standards and other regulations is crucial, and our products ensure that you achieve and maintain these essential benchmarks.
Protect Your Sensitive Processes and Products:
Maintaining a clean environment is crucial for developing pharmaceuticals or manufacturing semiconductors.
Minimize the risk of defects or failures in these sensitive processes for higher-quality outcomes and reduced product loss for your projects.