High-Tech Conversions Adhesive Floor Mats
Effectively capture dirt and dust from foot traffic and carts upon entrance into cleanroom
  • Stands up to heavy foot traffic and cart wheels Peel-off sheets eliminate need for messy cleaning Numbered corner tabs make tracking remaining sheets easy Adhesive backing ensures mats remain adhered to the floor
Available in various sizes and colors, these multilayer adhesive entrance floor mats are ideal for keeping dirt, dust, and contaminates from entering a cleanroom, laboratory, or other controlled environment. Each mat contains 30 fully coated particle-grabbing adhesive sheets, which easily peel off, revealing a new clean layer. A printed number in the corner of each low-density polyethylene (LDPE) sheet indicates how many layers remain and ensures at least one layer remains before replacement is needed.
$60.32 - $158.30CAD / Case of 4
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Width (in)
Availability Pricing
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 18" x 36", White; 4/cs RK-64100-51 Mfr # TT-1836-34W
COMPARE
White 36 18
$60.32CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 18" x 36", Gray; 4/cs RK-64100-52 Mfr # TT-1836-34G
COMPARE
Grey 36 18
$60.32CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 18" x 36", Blue; 4/cs RK-64100-53 Mfr # TT-1836-34B
COMPARE
Blue 36 18
$60.32CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 24" x 36", White; 4/cs RK-64100-54 Mfr # TT-2436-34W
COMPARE
White 36 24
$72.65CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 24" x 36", Blue; 4/cs RK-64100-55 Mfr # TT-2436-34B
COMPARE
Blue 36 24
$72.65CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 24" x 45", White; 4/cs RK-64100-56 Mfr # TT-2445-34W
COMPARE
White 45 24
$73.45CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 24" x 45", Blue; 4/cs RK-64100-57 Mfr # TT-2445-34B
COMPARE
Blue 45 24
$73.45CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 24" x 45", Gray; 4/cs RK-64100-58 Mfr # TT-2445-34G
COMPARE
Grey 45 24
$73.45CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 25" x 45", Gray; 4/cs RK-64100-59 Mfr # TT-2545-34G
COMPARE
Grey 45 25
$74.47CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 26" x 45", White; 4/cs RK-64100-60 Mfr # TT-2645-34W
COMPARE
White 45 26
$104.40CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 26" x 45", Blue; 4/cs RK-64100-61 Mfr # TT-2645-34B
COMPARE
Blue 45 26
$104.40CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 36" x 36", White; 4/cs RK-64100-62 Mfr # TT-3636-34W
COMPARE
White 36 36
$91.45CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 36" x 36", Gray; 4/cs RK-64100-63 Mfr # TT-3636-34G
COMPARE
Grey 36 36
$91.45CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 36" x 36", Blue; 4/cs RK-64100-64 Mfr # TT-3636-34B
COMPARE
Blue 36 36
$91.45CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 36" x 45", White; 4/cs RK-64100-65 Mfr # TT-3645-34W
COMPARE
White 45 36
$137.25CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 36" x 45", Blue; 4/cs RK-64100-67 Mfr # TT-3645-34B
COMPARE
Blue 45 36
$137.25CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 36" x 60", White; 4/cs RK-64100-68 Mfr # TT-3660-34W
COMPARE
White 60 36
$158.30CAD / Case of 4
High-Tech Conversions Adhesive Floor Mat with Multi-Layered Clean Films, 36" x 60", Blue; 4/cs RK-64100-69 Mfr # TT-3660-34B
COMPARE
Blue 60 36
$158.30CAD / Case of 4
Showing 1- 18 of 18